Skip to Content
Merck

L4385

α-Lactalbumin from bovine milk

Marker for non-denaturing PAGE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
235-747-3
NACRES:
NA.61
MDL number:
Biological source:
bovine milk
Form:
lyophilized powder
Technique(s):
microbiological culture: suitable
Concentration:
0.9—1.4 mg per vial protein (biuret)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Segment

form

lyophilized powder

mol wt

~14.2 kDa

concentration

0.9—1.4 mg per vial protein (biuret)

technique(s)

microbiological culture: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... LALBA(281894)

General description

α-Lactalbumin is a small, globular, whey protein that has been found in all milk studied to date. It is a metalloprotein of approximately 14 kDa produced in the mammary glands.

Application

α-Lactalbumin was used in the isolation and analysis of restriction endonuclease digestive patterns of chromosomal DNA from Mycobacterium paratuberculosis and other Mycobacterium species. It was also used in a study to test if the neuroendocrine protein 7B2 suppresses the aggregation of neurodegenerative disease-related proteins.

Biochem/physiol Actions

α-Lactalbumin is the cheif protein in human milk. It consists of a single polypeptide chain with 8 cysteines which form disulfide bridges. α-Lactalbumin binds several metal ions, including calcium, which is thought to play a role in the regeneration of native α-lactalbumin from the reduced, denatured form. α-Lactalbumin also has a distinct zinc binding site that is thought to play a role in the binding of the lactose synthase complex. The mature protein consists of 123 amino acid residues (14 kD), and it has a three-dimensional structure with 1.7 Α° resolution, demonstrating four α-helices and a triple stranded antiparallel β-sheet.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase. Site-directed mutagenesis of Asp87 or Asp88 to Ala completely abolishes the strong calcium binding affinity and reduces the stimulation of lactose synthase to <3.5% of the maximal rate.

Preparation Note

0.9-1.4 mg/mL after reconstitution with 1 mL of water


Still not finding the right product?

Explore all of our products under α-Lactalbumin from bovine milk


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library