Skip to Content
Merck
All Photos(1)

Key Documents

L3045

Sigma-Aldrich

β-Lactamase from Pseudomonas fluorescens

lyophilized powder, recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

lyophilized powder

enzyme activity

7.5-15 units/mg solid

shipped in

wet ice

storage temp.

2-8°C

Unit Definition

One unit will hydrolyze 1.0 μmole of benzylpenicillin per minute at pH 7.0 at 25 °C

Physical form

Lyophilized powder containing potassium phosphate and sodium bicarbonate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayuri Trivedi et al.
The Journal of the Association of Physicians of India, 60, 28-30 (2013-02-15)
In India where the prevalence of extended spectrum beta lactamase (ESBL) producing organisms among gram negative organisms is 60-70% and Ertapenem was unavailable at the beginning of this study, exclusive use of Group 2 Carbapenems (Imipenem and Meropenem) for treatment
A A Alsultan et al.
Journal of medical microbiology, 62(Pt 6), 885-888 (2013-03-23)
Carbapenem-resistant Acinetobacter baumannii is becoming increasingly prevalent in patients with diabetes mellitus in the Middle East. We examined the relationship of these bacteria and their resistance mechanisms to the diabetic disease status of patients in Saudi Arabia. Susceptibilities of 271
Jiri Klimes et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 44(1), 173-175 (2013-03-20)
Only a few reports exist on the occurrence of resistant bacteria in zoo animals. Therefore, an isolation of multiresistant Escherichia coli from the lungs of a captive South American tapir (Tapirus terrestris) lead to its characterization and further investigation of
Elena De Vecchi et al.
Journal of medical microbiology, 62(Pt 6), 859-863 (2013-03-12)
Urinary tract infections (UTIs) are a common cause of bacteraemia in the elderly and are associated with a high probability of hospitalization. Despite the impact of UTIs on health status and quality of life, a limited number of studies have
Molecular characterization of metallo-β-lactamase-producing Pseudomonas aeruginosa in a Czech hospital (2009-2011).
C C Papagiannitsis et al.
Journal of medical microbiology, 62(Pt 6), 945-947 (2013-03-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service