Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
NACRES:
NA.54
UNSPSC Code:
12352204
EC Number:
232-668-6
MDL number:
eCl@ss:
32160410
Specific activity:
≥200 units/mg protein
Biological source:
bovine milk
biological source
bovine milk
Quality Segment
form
lyophilized powder (essentially salt-free)
specific activity
≥200 units/mg protein
absorbance ratio
A412/280 nm 0.7-0.9
UniProt accession no.
storage temp.
−20°C
SMILES string
[O+H2]O[O-]
InChI
1S/H2O3/c1-3-2/h1-2H
InChI key
JSPLKZUTYZBBKA-UHFFFAOYSA-N
Gene Information
cow ... LPO(280844)
General description
Application
Lactoperoxidase from bovine milk has been used as a standard for milk lactoperoxidase to study the role of H2O2 in the inhibition of Staphylococcus aureus growth by Lactococcus garvieae in the presence of lactoperoxidase in raw milk. It has also been used in lactoperoxidase-mediated iodination to prepare radiolabeled peptides.
Biochem/physiol Actions
Lactoperoxidase contributes to the antimicrobial system of milk by inactivating a wide range of micro-organisms. This lactoperoxidase-mediated antimicrobial system is also identified in human secretions such as tear-fluid, saliva, and milk. Lactoperoxidase catalyzes the oxidation of molecules by releasing H2O2. The product exhibits antimicrobial activity.
Lactoperoxidase catalyzes the oxidation of iodide to iodine by hydrogen peroxide. This activity provides a gentle, specific alternative to chloramine T for the radioiodination of proteins and DNA.
Analysis Note
Protein determined by Lowry method.
Other Notes
One unit will oxidize 1.0 μmole of 2,2′-azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) at pH 5.5 at 25 °C.
Still not finding the right product?
Explore all of our products under Lactoperoxidase from bovine milk
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
