Skip to Content
Merck
All Photos(1)

Key Documents

L1920

Sigma-Aldrich

LY255283

≥98% (HPLC), powder

Synonym(s):

1-(5-Ethyl-2-hydroxy-4-(6-methyl-6-(1H-tetrazol-5-yl)heptyloxy)phenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28N4O3
CAS Number:
Molecular Weight:
360.45
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light yellow

solubility

DMSO: >20 mg/mL

originator

Eli Lilly

storage temp.

−20°C

SMILES string

CCc1cc(C(C)=O)c(O)cc1OCCCCCC(C)(C)c2nnn[nH]2

InChI

1S/C19H28N4O3/c1-5-14-11-15(13(2)24)16(25)12-17(14)26-10-8-6-7-9-19(3,4)18-20-22-23-21-18/h11-12,25H,5-10H2,1-4H3,(H,20,21,22,23)

InChI key

WCGXJPFHTHQNJL-UHFFFAOYSA-N

Application

LY255283, a competitive leukotriene B4 receptor antagonist, is used in leukotriene receptor research along with other selective leukotriene receptor agonists and antagonist to identify and differentiate the physiological and cell signaling effects of the leukotriene B4 receptor on processes such as paclitaxel resistance in MCF-7/DOX breast cancer cells, monocyte-derived dendritic cell chemotaxis, and 5-lipoxygenase activity and interleukin-8 production in human neutrophils. LY255283 may be used to help verify that an observed process or cell event is leukotriene B4 receptor-dependent.

Biochem/physiol Actions

LY255283 is a competitive leukotriene B4 receptor antagonist, with an IC50 of about 100 nM. It is somewhat selective for the BLT2 receptor, since IC50 values at the BLT1 receptor are >10 μM. LY255283 reduces airway obstruction in animal models of asthma.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Souza et al.
European journal of pharmacology, 403(1-2), 121-128 (2000-09-02)
Reperfusion of ischemic vascular beds may lead to recruitment and activation of leukocytes, release of mediators of the inflammatory process and further injury to the affected vascular bed and to remote sites. Neutrophils appear to play a major role in
Jin-Wook Lee et al.
The Journal of biological chemistry, 288(42), 30054-30063 (2013-08-30)
The majority of prostate cancer-related deaths are associated with advanced and metastatic malignancies. Although anoikis resistance has been recognized as one of the hallmarks of metastatic prostate malignancies, the molecular events that cause anoikis resistance are poorly understood. In this
M P Fink et al.
Critical care medicine, 21(12), 1825-1837 (1993-12-01)
To evaluate the hypothesis that treatment with LY255283, a novel leukotriene B4-receptor antagonist, is beneficial in an animal model of the adult respiratory distress syndrome induced by endotoxin. Prospective, randomized, controlled trial. Laboratory at a large university medical center. Twenty-five
J S Sawyer et al.
Journal of medicinal chemistry, 38(22), 4411-4432 (1995-10-27)
Structural derivatives of LY255283 have been studied as receptor antagonists of leukotriene B4. Substitution of the 2-hydroxyacetophenone subunit of 1 (LY255283) with a 2-arylphenol group provided entry into several new series that feature various mono- and diacidic core functionality. These
Eun-Young Kim et al.
Free radical biology & medicine, 49(6), 1072-1081 (2010-07-06)
Aggressive bladder cancer is a major cause of morbidity and mortality. Despite the fact that metastatic disease results in death in the majority of bladder cancer cases, the molecular events regulating the invasive phenotype of aggressive bladder cancer are not

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service