Skip to Content
Merck
All Photos(1)

Key Documents

L121

Sigma-Aldrich

Levallorphan tartrate salt

≥98% (HPLC), powder

Synonym(s):

17-(2-Propenyl)morphinan-3-ol tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NO · C4H6O6
CAS Number:
Molecular Weight:
433.49
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]22/D −35°, c = 0.38 in H2O(lit.)

color

white to beige

solubility

H2O: 5 mg/mL, clear

originator

Roche

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.[H][C@@]12CCCC[C@@]13CCN(CC=C)[C@@H]2Cc4ccc(O)cc34

InChI

1S/C19H25NO.C4H6O6/c1-2-10-20-11-9-19-8-4-3-5-16(19)18(20)12-14-6-7-15(21)13-17(14)19;5-1(3(7)8)2(6)4(9)10/h2,6-7,13,16,18,21H,1,3-5,8-12H2;1-2,5-6H,(H,7,8)(H,9,10)/t16-,18+,19+;1-,2-/m01/s1

InChI key

FWMLYVACGDQRFU-ZTMWJVNESA-N

Gene Information

Biochem/physiol Actions

Partial agonist (antagonist) at μ and δ opioid receptors.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Photosensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Chen Chen et al.
International journal of nanomedicine, 10, 7265-7274 (2015-12-15)
To alleviate the inherent problems of amphotericin B (AmB), such as poor water solubility and nephrotoxicity, a novel self-assembling mixed polymeric micelle delivery system based on lecithin and combined with amphiphilic polymers, Pluronic(®), Kolliphor(®), d-alpha tocopheryl polyethylene glycol succinate, and
Tzu-Pin Li et al.
Scientific reports, 7(1), 10674-10674 (2017-09-08)
This study involved physical and pharmacokinetic characterizations of trans-resveratrol (t-Rev)-loaded saLMPMs which attempted to improve t-Rev's pharmacokinetic profiles and bioavailability resolving hurdles limiting its potential health benefits. The optimal formulation consisted of t-Rev, lecithin, and Pluronic
Ghada A Abdelbary et al.
Drug delivery, 24(1), 309-319 (2017-02-07)
Vesicular drug carriers for ocular delivery have gained a real potential. Proniosomal gels as ocular drug carriers have been proven to be an effective way to improve bioavailability and patient compliance. Formulation and in vitro/ex vivo/in vivo characterization of ketoconazole
Anushree Seth et al.
International journal of nanomedicine, 7, 5129-5136 (2012-10-12)
The influence of morphology and surface properties on the therapeutic efficacy of degradable polymeric microparticles has not been well understood. One of the primary reasons for this is the limited ability to fabricate microparticles with controlled morphology and surface properties.
Ling-Chun Chen et al.
International journal of nanomedicine, 11, 1557-1566 (2016-05-05)
Quercetin (Que) is known to have biological benefits including an anticancer effect, but low water solubility limits its clinical application. The aim of this study was to develop a lecithin-based mixed polymeric micelle (LMPM) delivery system to improve the solubility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service