Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C7H16N2O2 · 2HCl
CAS Number:
Molecular Weight:
233.14
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
247-625-7
MDL number:
Pricing and availability is not currently available.
Product Name
L-Lysine methyl ester dihydrochloride,
assay
≥99% (TLC)
Quality Segment
form
powder
color
white
storage temp.
−20°C
SMILES string
Cl.COC(=O)C(N)CCCCN
InChI
1S/C7H16N2O2.ClH/c1-11-7(10)6(9)4-2-3-5-8;/h6H,2-5,8-9H2,1H3;1H
InChI key
FORVAIDSGSLRPX-UHFFFAOYSA-N
Application
L-Lysine methyl ester is used in the production of lysine methyl ester based cationic surfactants and hydrogels.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description ≥99.0% (AT) | description certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | description ≥98%, FG |
| form powder | form solid | form - | form - |
| assay ≥99% (TLC) | assay ≥99.0% (AT) | assay - | assay ≥98% |
| storage temp. −20°C | storage temp. - | storage temp. - | storage temp. - |
| Quality Level 200 | Quality Level 100 | Quality Level 300 | Quality Level 300 |
| color white | color - | color white | color - |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



