Skip to Content
Merck
All Photos(2)

Key Documents

L0159

Sigma-Aldrich

Anti-Luciferase antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti Luciferase Antibody, Firefly Luciferase Antibody, Luciferase Antibody, Luciferase Antibody - Anti-Luciferase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

packaging

antibody small pack of 25 μL

technique(s)

indirect immunofluorescence: 1:500-1:1,000 using eukaryotic cells transfected with a plasmid bearing the luciferase gene
western blot: 1:10,000 using purified Luciferase

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Analysis of gene expression is commonly assayed by transient transfection. These systems are usually based on the use of fusion genes which are inserted into cells, and expression of the gene is assayed within 48 hours after introduction of DNA. Usually the fusion consists of the promoter binding site or enhancer sequence under study which is attached to a reporter gene. The amount of the reporter protein synthesized under the experimental conditions is presumed to reflect the ability of the sequences studied to direct or promote transcription.
Luciferase has become one of the more widely used reporter enzymes. The reporter plasmid contains the gene from the firefly Photinus pyralis. The enzyme catalyzes a bioluminescent reaction which requires the substrate luciferin as well as Mg+2 and ATP. Mixing these reagents with the cell extract containing luciferase results in a flash of light that decays rapidly. This light can be detected by a luminometer. The total light emission is proportional to the luciferase activity of the sample. The luciferase assay is fast and sensitive and does not require a radioactive substrate as in the CAT assay. A disadvantage of the luciferase assay is that it requires a rather expensive instrument, the luminometer, to measure enzyme activity.

Specificity

The antibody identifies recombinant luciferase in eukaryotic cells transfected with a plasmid bearing the luciferase gene.

Immunogen

firefly (Photinus pyralis) luciferase.

Application

Anti-Luciferase antibody produced in rabbit has been used in immunoblotting and immunohistochemistry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marco Quarta et al.
Nature communications, 8, 15613-15613 (2017-06-21)
Volumetric muscle loss (VML) is associated with loss of skeletal muscle function, and current treatments show limited efficacy. Here we show that bioconstructs suffused with genetically-labelled muscle stem cells (MuSCs) and other muscle resident cells (MRCs) are effective to treat
A rapid and quantitative coat protein complex II vesicle formation assay using luciferase reporters
Fromme J Chris and Kim J
Analytical biochemistry, 421(2), 482-488 (2012)
Yuying Sang et al.
Plant communications, 1(4), 100025-100025 (2020-12-29)
Effector proteins delivered inside plant cells are powerful weapons for bacterial pathogens, but this exposes the pathogen to potential recognition by the plant immune system. Therefore, the effector repertoire of a given pathogen must be balanced for a successful infection.
Intraductal delivery of adenoviruses targets pancreatic tumors in transgenic Ela-myc mice and orthotopic xenografts
Jose A, et al.
Testing, 4(1), 94-94 (2013)
Marco Chiabudini et al.
Molecular and cellular biology, 32(23), 4769-4779 (2012-09-26)
When a polyadenylated nonstop transcript is fully translated, a complex consisting of the ribosome, the nonstop mRNA, and the C-terminally polylysine-tagged protein is generated. In Saccharomyces cerevisiae, a 3-step quality control system prevents formation of such dead-end complexes. Nonstop mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service