Skip to Content
Merck

K7250

Kidney acetone powder porcine

Type II

Synonym(s):

Porcine Kidney Acetone Extract

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.26
MDL number:
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type II

Quality Level

storage temp.

−20°C

Application

Kidney acetone powder porcine has been used to hydrolyze folates.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Nutritional characterization of traditional and improved dihe,
alimentary blue-green algae from the lake Chad region in Africa
Marina Carcea
Food Sci. Technol., 753-763 (2015)
Study of wheat breakfast rolls fortified
with folic acid
European Journal of Nutrition, 279-286 (2002)



Global Trade Item Number

SKUGTIN
K7250-10G04061832408682