Skip to Content
Merck

K6250

2-Keto-D-gluconic acid hemicalcium salt hydrate

Synonym(s):

D-Arabino-2-hexulopyranosonic acid hemicalcium salt hydrate, Calcium 2-keto-D-gluconate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H9O7 · ½Ca · xH2O
CAS Number:
Molecular Weight:
213.17 (anhydrous basis)
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3808734
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (TLC)

Quality Segment

form

powder

technique(s)

thin layer chromatography (TLC): suitable

solubility

water: 49-51 mg/mL

storage temp.

2-8°C

SMILES string

[H]O[H].OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)C(=O)O[Ca]OC(=O)C(=O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H10O7.Ca.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-4,7-10H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+;;/m11../s1

InChI key

LBTJSFDOKRLTMQ-LWDDDOIUSA-L

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves

target_organs

Respiratory system

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library