K4003
Knudson C Modified Orchid Medium
powder, suitable for plant cell culture
Synonym(s):
KC Orchid Media, modified
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.72
Recommended Products
form
powder
Quality Level
technique(s)
cell culture | plant: suitable
application(s)
agriculture
storage temp.
2-8°C
Application
Knudson C Modified Orchid Medium is a nutrient solution for Orchid (Orchidaceae) seed germination. This media is frequently supplemented with 100-150 g banana, or 100-200 mL coconut water, and 2 g of vegetable charcoal per liter.
Formula variant
With the macro- and micronutrients as described by Knudson (1946). Contains sucrose.
Media Formulation
Media Formulation
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Philip J Kauth et al.
Annals of botany, 102(5), 783-793 (2008-09-02)
Ecotypic differentiation has been explored in numerous plant species, but has been largely ignored in the Orchidaceae. Applying a specific germination protocol for widespread seed sources may be unreliable due to inherent physiological or genetic differences in localized populations. It
E Ouni et al.
Human reproduction (Oxford, England), 35(6), 1391-1410 (2020-06-17)
How do elastic matrisome components change during the lifetime of the human ovary? The deposition and remodeling of mechanical matrisome components (collagen, elastin, elastin microfibril interface-located protein 1 (EMILIN-1), fibrillin-1 and glycosaminoglycans (GAGs)) that play key roles in signaling pathways
Guek Eng Sim et al.
Plant cell reports, 26(4), 383-393 (2006-10-07)
We have successfully developed a method to induce early in vitro flowering of the self-pollinated seedlings of a tropical orchid hybrid, Dendrobium Madame Thong-In. Transition of vegetative shoot apical meristem to inflorescence meristem was observed when young protocorms were cultured
Kate Lykke Lambertsen et al.
PloS one, 7(10), e47744-e47744 (2012-10-19)
The calmodulin/calcium-activated K(+) channel KCa3.1 is expressed in red and white blood cells, epithelia and endothelia, and possibly central and peripheral neurons. However, our knowledge about its contribution to neurological functions and behavior is incomplete. Here, we investigated whether genetic
Giovanni Di Liberto et al.
Cell, 175(2), 458-471 (2018-09-04)
Inflammatory disorders of the CNS are frequently accompanied by synaptic loss, which is thought to involve phagocytic microglia and complement components. However, the mechanisms accounting for aberrant synaptic connectivity in the context of CD8+ T cell-driven neuronal damage are poorly understood.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service