Skip to Content
Merck

K2252

Diethyl β-ketoadipate

Synonym(s):

β-Ketoadipic acid diethyl ester, 3-Oxohexanedioic acid diethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H16O5
CAS Number:
Molecular Weight:
216.23
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
230-479-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

storage temp.

−20°C

SMILES string

CCOC(=O)CCC(=O)CC(=O)OCC

InChI

1S/C10H16O5/c1-3-14-9(12)6-5-8(11)7-10(13)15-4-2/h3-7H2,1-2H3

InChI key

ONRIYMIXCUUSNS-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library