Skip to Content
Merck
All Photos(1)

Key Documents

K1757

Sigma-Aldrich

Keratinocyte Growth Factor human

≥95% (SDS-PAGE and HPLC), recombinant, expressed in E. coli, powder, suitable for cell culture

Synonym(s):

hFGF-7, hKGF, FGF-7, KGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Keratinocyte Growth Factor human, KGF, recombinant, expressed in E. coli, powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE and HPLC)

form

powder

potency

0.1-15 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

18.9 kDa

packaging

pkg of 10 and 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF7(2252)

Looking for similar products? Visit Product Comparison Guide

General description

Keratinocyte Growth Factor(KGF) is a 18.9 kDa growth factor, spanning 163 amino acids, belongs to the fibroblast growth factor family. It includes a secretory signal sequence and shares 39% identity with FGF-Basic. It is expressed in a number of stromal fibroblast cell lines, mesenchymal cells but absent from normal glial cells and epithelial cell lines.

Application

Keratinocyte Growth Factor human has been used:
  • as a component of epithelial growth medium
  • as a positive control for antigen to determine the activity of the antibody
  • to analyze the influence of KGF associated with thermo-sterilized carboxymethyl chitosan (NOCCts) in the reduction of pericardial adhesions

Biochem/physiol Actions

Keratinocyte Growth Factor(KGF) stimulates proliferation and differentiation of keratinocytes and other cells in the epithelium. It induces chemotaxis of microvascular endothelial cells and neovascularization of rat cornea. It stimulates the hyaluronic acid synthesis in proliferating and migrating basal cells. KGF contributes to the survival of cells under stress conditions. KGF is highly expressed in injured and inflamed epithelial cells, thereby, promotes wound healing.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.5 mg bovine serum albumin.

Analysis Note

Determined by a cell proliferation assay using 4MBr-5 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristics of MIC-1 antlerogenic stem cells and their effect on hair growth in rabbits.
Cegielski M, et al.
In Vivo, 27(1), 97-106 (2013)
Keratinocyte growth factor (KGF) is required for postnatal thymic regeneration
Alpdogan O, et al.
Blood, 107(6), 2453-2460 (2006)
P Gillis et al.
Journal of cell science, 112 ( Pt 12), 2049-2057 (1999-05-26)
Keratinocyte growth factor (KGF), also called fibroblast growth factor-7, is widely known as a paracrine growth and differentiation factor that is produced by mesenchymal cells and has been thought to act specifically on epithelial cells. Here it is shown to
Stimulation of in vivo angiogenesis by in situ crosslinked, dual growth factor-loaded, glycosaminoglycan hydrogels
Elia R, et al.
Biomaterials, 31(17), 4630-4638 (2010)
Synergism between keratinocyte growth factor and carboxymethyl chitosan reduces pericardial adhesions
Lopes JB, et al.
The Annals of Thoracic Surgery, 90(2), 566-572 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service