Skip to Content
Merck
All Photos(1)

Key Documents

K113

Sigma-Aldrich

Ketamine hydrochloride/xylazine hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

solution

Quality Level

drug control

USDEA Schedule III; stupéfiant (France); regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain)

color

colorless

storage temp.

2-8°C

SMILES string

Cl.Cl.Cc1cccc(C)c1NC2=NCCCS2.CNC3(CCCCC3=O)c4ccccc4Cl

InChI

1S/C13H16ClNO.C12H16N2S.2ClH/c1-15-13(9-5-4-8-12(13)16)10-6-2-3-7-11(10)14;1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12;;/h2-3,6-7,15H,4-5,8-9H2,1H3;3,5-6H,4,7-8H2,1-2H3,(H,13,14);2*1H

InChI key

IYJHJWJNWIPQFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

This is a solution of ketamine hydrochloride, an NMDA receptor antagonist, and xylazine hydrochloride, an α2- adrenergic receptor agonist.

Other Notes

Telephone orders are not accepted for this product.

Kit Components Only

Product No.
Description

  • Ketamine hydrochloride 80 mg/mL

  • Xylazine hydrochloride 12 mg/mL

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Tepper et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 15(4), 3092-3103 (1995-04-01)
Evidence from electrophysiological studies has suggested an inhibitory interaction between GABAergic neurons in substantia nigra pars reticulata and dopaminergic neurons in pars compacta. However, that this inhibitory interaction is due to a projection from pars reticulata to pars compacta has
Yuanlin He et al.
Cell death & disease, 8(5), e2817-e2817 (2017-05-26)
In mammalian ovaries, primordial follicles remain in a quiescent state until activation by the surrounding microenvironment. Ovarian intervention, for example, ovarian cystectomy, ovarian wedge resection or laser drilling therapies for polycystic ovarian syndrome, has long been reported to change follicular
Liyuan Jiang et al.
Aging and disease, 11(3), 603-617 (2020-06-04)
The complex pathology of chronic thoracic spinal cord compression involves vascular and neuroarchitectural repair processes that are still largely unknown. In this study, we used synchrotron radiation microtomography (SRμCT) to quantitatively characterize the 3D temporal-spatial changes in the vascular and
J P Gratton et al.
British journal of pharmacology, 114(3), 720-726 (1995-02-01)
1. In the present study, the precursors of endothelin-1, endothelin-2 and endothelin-3 were tested for their pressor and bronchoconstrictor properties in the anaesthetized guinea-pig. In addition, the effects of big-endothelin-1 and endothelin-1 were assessed under urethane or ketamine/xylazine anaesthesia. 2.
Azim Ullah Shamsul Islam et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-04)
Physiological Glucocorticoids are important regulators of the immune system. Pharmacological GCs are in widespread use to treat inflammatory diseases. Adrenalectomy (ADX) has been shown to exacerbate renal injury through inflammation and oxidative stress that results in renal impairment due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service