Skip to Content
Merck

J2000

Juvenile hormone III

≥65%, liquid, non-sterile

Synonym(s):

Methyl farnesoate, 10,11-epoxide, C16-Juvenile Hormone, JH-III, Manduca hormone, trans,trans-10,11-Epoxy-3,7,11-trimethyl-2,6-dodecadienic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
UNSPSC Code:
51111800
NACRES:
NA.32
PubChem Substance ID:
EC Number:
246-072-9
Beilstein/REAXYS Number:
1316317
MDL number:
Biological source:
synthetic (organic)
Form:
liquid
Assay:
≥65% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Juvenile hormone III, ≥65%, liquid, non-sterile

InChI key

QVJMXSGZTCGLHZ-ZPLWXOMKSA-N

InChI

1S/C16H26O3/c1-12(9-10-14-16(3,4)19-14)7-6-8-13(2)11-15(17)18-5/h7,11,14H,6,8-10H2,1-5H3/b12-7+,13-11+

SMILES string

COC(=O)\C=C(/C)CC\C=C(/C)CC[C@H]1OC1(C)C

biological source

synthetic (organic)

sterility

non-sterile

assay

≥65% (HPLC)

form

liquid

concentration

≥65%

technique(s)

activity assay: suitable

shipped in

ambient

storage temp.

−20°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

Application

Juvenile hormone III has been used to:
  • study the effect of juvenile hormone on mictic (sexual) female production of the rotifer Brachionus plicatilis Muller
  • study the effect of juvenile hormone on head GB19811 (putative Takeout/juvenile hormone binding protein) mRNA levels in adult honeybees
  • study the effect of juvenile hormone on gonadotropic and physiological functions in bumblebee Bombus terrestris

Biochem/physiol Actions

Controls the larval metamorphosis of insects, including the retention of juvenile characteristics.
JHBPs (JH-binding proteins) protect JH (juvenile hormone) from JH esterase- and epoxide hydrolase-mediated degradation. They also help in delivering JH to target tissues.Juvenile hormones(JHs) are a group of acyclic sesquiterpenoids, and in some insect lineages, the farnesol backbone of these compounds undergoes chemical modification to produce a homologous series of hormones. JH is essential for insect development and reproduction in every aspect. It plays a key role in metamorphosis and caste determination in social insects. It modulates behavior, polyphenism, larval and adult diapause, and ovarian development. The sesquiterpenoid backbone of juvenile hormone III (JH III) is biosynthesized through the classical mevalonate (MVA) pathway in cockroaches.

General description

Research Area: Cell Signaling

Juvenile hormone III (JH III) is the most prevalent juvenile hormone (JH) found in insects.

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linquan Ge et al.
Frontiers in physiology, 11, 579233-579233 (2020-10-13)
The mirid bug, Cyrtorhinus lividipennis Reuter, is an important predator of rice planthoppers in Asia. In a previous study, C. lividipennis fed on gramineous weeds with brown planthopper (BPH) eggs had reduced development compared to those fed on rice with
Toyomi Kotaki et al.
Journal of insect physiology, 57(1), 147-152 (2010-10-26)
Juvenile hormone III skipped bisepoxide (JHSB(3)), methyl (2R,3S,10R)-2,3;10,11-bisepoxyfarnesoate was recently determined as a novel juvenile hormone (JH) in a stink bug, Plautia stali. To further confirm the biological function of JHSB(3) in this insect, its juvenilizing, reproduction-stimulating and diapause-terminating activities
Atul Pandey et al.
Neurobiology of sleep and circadian rhythms, 9, 100056-100056 (2020-12-29)
The circadian and endocrine systems influence many physiological processes in animals, but little is known on the ways they interact in insects. We tested the hypothesis that juvenile hormone (JH) influences circadian rhythms in the social bumble bee Bombus terrestris.
Keiji Matsumoto et al.
Royal Society open science, 8(2), 202242-202242 (2021-05-12)
Juvenile hormone (JH) plays important roles in almost every aspect of insect development and reproduction. JHs are a group of acyclic sesquiterpenoids, and their farnesol backbone has been chemically modified to generate a homologous series of hormones in some insect
Jesus Lozano et al.
Scientific reports, 1, 163-163 (2012-02-23)
Insect metamorphosis is regulated by ecdysteroids, which induce molts, and juvenile hormone (JH), which inhibits metamorphic changes. The molecular action of ecdysteroids has been thoroughly studied, but that of JH is poorly understood, with data currently only being available for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service