Skip to Content
Merck
All Photos(1)

Key Documents

ISO2

Sigma-Aldrich

Mouse Monoclonal Antibody Isotyping Reagents

sufficient for 1000 tests (clones) (by ELISA), sufficient for 40 tests (clones) (by immunodiffusion, ODD)

Synonym(s):

Mouse Antibody Isotyping Kit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.32

usage

sufficient for 1000 tests (clones) (by ELISA)
sufficient for 40 tests (clones) (by immunodiffusion, ODD)

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The Mouse Monoclonal Antibody Isotyping Reagents enable qualitative isotype determination of mouse monoclonal antibosies derived from hybridoma supernatant, ascites fluid or purified forms.

Specificity

ISO-2 contains 0.2 mL vials of subclass specific antibodies to all four mouse IgG subclasses, IgA, and IgM. Directions are included for using these reagents in ELISA and Ouchterlony assays to determine the subclass of monoclonal antibodies in ascites, culture supernatants, or purified preparations. Additional reagents may be required, such as conjugates for ELISA assays or agarose and buffer components for Ouchterlony assays.

Application

Determination of the subclass of a monoclonal antibody is helpful for characterization of the antibody, for choosing detection reagents, and for deciding on a purification scheme.
Mouse Monoclonal Antibody Isotyping Reagents has been used in monoclonal antibody isotype determination using enzyme-linked immunosorbent assay (ELISA). It may be used in Ouchterlony immunodiffusion assays.

Features and Benefits

  • May be used in a variety of assay formats
  • Suitable for all antibody forms
  • Determines all mouse IgG subclasses, IgA, and IgM

Kit Components Only

Product No.
Description

  • goat antisera to mouse IgA .2 mL

  • goat antisera to mouse IgG3 .2 mL

  • goat antisera to mouse IgG1 .2 mL

  • goat antisera to mouse IgG2a .2 mL

  • goat antisera to mouse IgG2b .2 mL

  • goat antisera to mouse IgM .2 mL

related product

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorma Hinkula et al.
Vaccines, 7(3) (2019-07-19)
Background: Vaccination is commonly used to prevent and control influenza infection in humans. However, improvements in the ease of delivery and strength of immunogenicity could markedly improve herd immunity. The aim of this pre-clinical study is to test the potential
Selvaraj Pavulraj et al.
Veterinary microbiology, 210, 188-196 (2017-11-07)
Equine influenza viruses (EIVs) are responsible for acute contagious respiratory infection in equines and the disease remains a major threat for equine population throughout the world despite vaccination strategies in place. The present study was aimed to assess the suitability
Violetta Sączyńska et al.
Virology journal, 15(1), 13-13 (2018-01-18)
The highly pathogenic avian influenza viruses of the H5 subtype, such as the H5N1 viral strains or the novel H5N8 and H5N2 reassortants, are of both veterinary and public health concern worldwide. To combat these viruses, monoclonal antibodies (mAbs) against
Generation and characterization of novel stromal specific antibodies
Halder S, et al.
Cell Research, 15(9), 739-739 (2005)
Qimin Wang et al.
Frontiers in microbiology, 11, 1961-1961 (2020-09-15)
The extracellular domain of influenza M2 protein (M2e) is highly conserved and is a promising target for development of universal influenza vaccines. Here, we synthesized a peptide vaccine consisting of M2e epitope linked to a fibrillizing peptide, which could self-assemble

Protocols

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Detailed ELISA protocols cover indirect and capture ELISA techniques, recommending products for sandwich and indirect ELISA experiments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service