Skip to Content
Merck
All Photos(1)

Key Documents

I7276

Sigma-Aldrich

Anti-Insulin-Like Growth Factor-II antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IGF-II

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5-15 μg/mL using human Placenta
neutralization: 0.2-0.8 μg/mL
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGF2(3481)

General description

Insulin-like growth factors -I and -II are important regulators of signalling that mediates growth and metabolism in cells. Insulin-like growth factor -II (IGF-II) is widely expressed during embryonic stages and regulates the placental development. IGF-II acts via specific receptor, IGF-IIR, the binding does not result in signal transduction, but is a mechanism to restrict the bioavailability of IGF-II. IGF-II also binds to IGF-IR which results in the activation of MAPK, Raf/Ras and PI3K cascades. The effects of IGF-II signalling are important in growth of blood vessels (angiogenesis) gene transcription, proliferation and transformation. Irregular IGF-II signaling has been implicated in many cancers including lung cancer, colo-rectal cancer, cervical cancer, testicular cancer, rhabdosarcoma, melanoma and hepatoblastoma
Anti-IGF-II recognizes human insulin-like growth factor II. It does not react with human IGF-I.

Specificity

The antibody will neutralize the biological activity of recombinant human IGF-II. It does not neutralize the biological activity of recombinant human IGF-I.

Immunogen

recombinant human IGF-II

Application

Anti- IGF-II antibody may be used for neutralization reactions, the ND50 of the antibody is 0.2-0.8 μg/ml. This antibody was used to neutralize IGF-II in glioblastoma cell lines. For immunohistochemistry a working antibody concentration of 5-15 μg/ml may be used for formalin-fixed, paraffin-embedded human placenta sections. The antibody is suitable for immunoblotting.

Physical form

Lyophilized from a 0.2 μm filtered solution of phosphate buffered saline, pH 7.4, with 5% trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonino Belfiore et al.
Endocrine reviews, 30(6), 586-623 (2009-09-16)
In mammals, the insulin receptor (IR) gene has acquired an additional exon, exon 11. This exon may be skipped in a developmental and tissue-specific manner. The IR, therefore, occurs in two isoforms (exon 11 minus IR-A and exon 11 plus
Ramaswamy Suvasini et al.
The Journal of biological chemistry, 286(29), 25882-25890 (2011-05-27)
Glioblastoma is the most common and malignant form of primary astrocytoma. Upon investigation of the insulin-like growth factor (IGF) pathway, we found the IGF2BP3/IMP3 transcript and protein to be up-regulated in GBMs but not in lower grade astrocytomas (p <
Wendy Chao et al.
Cytokine & growth factor reviews, 19(2), 111-120 (2008-03-01)
Insulin-like growth factor II (IGF2) is perhaps the most intricately regulated of all growth factors characterized to date. Its gene is imprinted--only one allele is active, depending on parental origin--and this pattern of expression is maintained epigenetically in almost all
Robert G Maki
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 28(33), 4985-4995 (2010-10-27)
Insulin-like growth factors were discovered more than 50 years ago as mediators of growth hormone that effect growth and differentiation of bone and skeletal muscle. Interest of the role of insulin-like growth factors in cancer reached a peak in the
O Manousos et al.
International journal of cancer, 83(1), 15-17 (1999-08-17)
Recent data suggest that the IGF system plays an important role in the pathogenesis of several forms of human cancer, and there is evidence that IGFs acting in an autocrine and paracrine manner may also affect colorectal cancer risk. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service