Skip to Content
Merck

I7016

H-7 dihydrochloride

≥98% (HPLC)

Synonym(s):

1-(5-Isoquinolinylsulfonyl)-2-methylpiperazine dihydrochloride, Isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H17N3O2S · 2HCl
CAS Number:
Molecular Weight:
364.29
UNSPSC Code:
12352200
NACRES:
NA.77
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5840763
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

solubility

H2O: 20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Cl.CC1CNCCN1S(=O)(=O)c2cccc3cnccc23

InChI

1S/C14H17N3O2S.2ClH/c1-11-9-16-7-8-17(11)20(18,19)14-4-2-3-12-10-15-6-5-13(12)14;;/h2-6,10-11,16H,7-9H2,1H3;2*1H

InChI key

OARGPFMFRLLKPF-UHFFFAOYSA-N

General description

H-7 dihydrochloride blocks human immunodeficiency virus (HIV-1) replication in MOLT-4 (clone No. 8) cell line. It increases the secretion of interleukin 1β (IL-1β).

Application

H-7 dihydrochloride has been used to study H-7-induced inhibition of contractility in rat embryo fibroblasts (REF52) cells and acts as a kinase inhibitor.

Biochem/physiol Actions

Inhibitor of cyclic nucleotide dependent protein kinase (PKA) and protein kinase C.

Disclaimer

Light sensitive


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library