Skip to Content

I6160

ITK (352-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

EMT, LYK, MG C126257, MGC126258, PSCTK2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Form:
buffered aqueous glycerol solution
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells
Mol wt:
~53 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

49-67 nmol/min·mg

mol wt

~53 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ITK(3702)

General description

ITK (IL2-inducible T-cell kinase) is a member of the Tec family of tyrosine kinases. ITK is expressed in T-cells.

Biochem/physiol Actions

ITK (IL2-inducible T-cell kinase) is important for T-cell activation through the T-cell antigen receptor. Absence of ITK in T cells causes impairment in TCR (T cell receptor)-mediated actin polymerization, cell polarization and signaling events associated with cytoskeletal reorganization.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library