Skip to Content
Merck

Skip To

I6138

Idazoxan hydrochloride

α2-adrenoceptor antagonist

Synonym(s):

(±)-2-[1,4-Benzodioxan-2-yl]-2-imidazoline hydrochloride, RX 781094

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H12N2O2 · HCl
CAS Number:
Molecular Weight:
240.69
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Idazoxan hydrochloride,

Quality Segment

solubility

H2O: 50 mg/mL

SMILES string

Cl.C1CN=C(N1)C2COc3ccccc3O2

InChI

1S/C11H12N2O2.ClH/c1-2-4-9-8(3-1)14-7-10(15-9)11-12-5-6-13-11;/h1-4,10H,5-7H2,(H,12,13);1H

InChI key

MYUBYOVCLMEAOH-UHFFFAOYSA-N

Gene Information

Application

Idazoxan hydrochloride has been used to study the efficacy of antidepressant treatments that interact with multiple neurotransmitter systems.

Biochem/physiol Actions

α2-adrenoceptor antagonist; I2 imidazoline receptor agonist; I1 imidazoline receptor antagonist. Idazoxan can antagonize various behaviors generated by ethanol in a preclinical setting. It possesses neuroprotective activity against spinal cord injury, resulted due to experimental autoimmune encephalomyelitis (EAE) in mouse, an animal modal of multiple sclerosis (MS).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R9525A0779C7897
description

-

description

solid, ≥98% (HPLC)

description

solid

description

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

solubility

H2O: 50 mg/mL

solubility

H2O: >20 mg/mL

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1.4 mg/mL (Solutions may be stored for several days at 4 °C), boiling water: 100 mg/mL (Solutions may be stored for several days at 4 °C), 0.1 M HCl: soluble (Solutions may be stored for several days at 4 °C), methanol: soluble (Solutions may be stored for several days at 4 °C)

solubility

H2O: soluble, methanol: soluble

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters