Skip to Content

I4777

Interferon-γ from mouse

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IFN-γ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

protein 15.6 kDa

packaging

pkg of 0.1 mg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

solubility

water: soluble 1.000 mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Ifng(15978)

Application

Interferon-γ from mouse has been used:
  • for in vitro stimulation of the BMDC (bone marrow derived dendritic cell), BMM (bone marrow-derived macrophages) and CD11c+ (CD11 antigen-like family member C) lung macrophages
  • for stimulation of murine macrophages to induce the production of nitrite and nitrate
  • to check CCAAT/enhancer binding protein β (C/EBPβ)-mediated changes in the expression of prostaglandin synthesis enzymes in neuroinflammation

Biochem/physiol Actions

Ifng (interferon-γ) plays an important role in innate and adaptive immune responses, and helps in defense against bacterial infections, dimorphic yeast and some viral infections.

Physical form

Lyophilized from a 0.2 μm filtered buffered solution. Package size based on protein content.

Analysis Note

The biological activity of recombinant mouse IFN-γ is measured by its ability to inhibit the proliferation of murine WEHI-279 cells.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library