Skip to Content
Merck

I4765

Human IgG4 Lambda, Recombinant Protein

purified immunoglobulin

Synonym(s):

Human IgG4 Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Biological source:
human (recombinant)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human (recombinant)

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

shipped in

dry ice

storage temp.

−20°C

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. IgG4 is the least abundant of the IgG subclass. IgG4-related systemic diseases include autoimmune pancreatitis, mediastinal fibrosis and multiple sclerosis.
IgG4, λ recombinant protein is obtained by expression of a phage library in a human cell line and purified by affinity chromatography.

Application

The purified IgG4, λ may be used as an immunoglobulin calibrator, reference antigen, blocking agent, or coating protein in a variety of immunoassays including ELISA, dot blot immunobinding, Western immunobloting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell binding assays. Purified IgG4, λ was used as isotype control in flow cytometry of chronic lymphocytic leukemia and mantle cell lymphoma cells. It was also used for lectin- surface plasmon resonance and IgG subclass labelling by flow cytometry at a concentration of 50 μg/mL.

Analysis Note

The purity and identity are determined by immunoelectrophoresis, indirect ELISA, and SDS-PAGE.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library