Skip to Content
Merck

Skip To

HT90T

Gram Stain Kit for Tissue

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
41116121
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

Application

Tissues are stained for Gram-positive organisms with crystal violet, mordanted in iodine, decolorized, stained for Gram-negative organisms with safranin O and then counterstained with tartrazine. For delineating Gram-positive and Gram-negative organisms in paraffin embedded tissues.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HT90AR4132HT10316
technique(s)

microbe id | staining: suitable

technique(s)

-

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

form

solution

form

solution

form

solution

form

solution

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.


Kit Components Only

Product No.
Description

  • Crystal Violet Solution (HT90-1) 8 fl oz

  • Gram's Iodine Solution (HT90-2) 8 fl oz

  • Safranin O Solution (HT90-4) 8 fl oz

  • Tartrazine Solution (HT30-2-8) 250 mL

signalword

Danger

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardCorrosion

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. How many slides can be stained with the HT90T-1KT kit?

    1 answer
    1. The instructions in the HT90T kit advise to "flood" the slide with the various reagents provided. Each bottle contains 8 fluid ounces or approximately 237 ml. If 1 ml is used per slide, one can expect a maximum of 237 slides per kit. Using more reagent could reduce this number, potentially down to about 120 slides per kit. The thickness or size of the specimen placed on the slide may also impact reagent usage. The volume of differentiator used can vary depending on the specimen and the individual performing the assay.

      A realistic number is likely in the range of 100 to 237 slides. However, this should be considered a rough estimate, not an absolute guarantee.

      The HT90T kit is recommended for staining tissue sections attached to glass slides. In contrast, the HT90A kit is intended for staining smear prepared on glass slides. For reagent usage, the two different kits should stain an equal number of slides or specimens.

      Helpful?

Reviews

No rating value

Active Filters