Skip to Content
Merck

Skip To

HT902

Gram′s Iodine Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
41116121
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Application

Used as a kit component in Gram Stain Procedure (Sigma Procedure HT90).

Other Notes

Iodine, 0.33%, and potassium iodide, 0.66%.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HT902322852HT254
application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

form

solution

form

solution

form

solution

form

solution

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

IVD

for in vitro diagnostic use

shelf life

Expiry date on the label

shelf life

Expiry date on the label

shelf life

Expiry date on the label.

shelf life

Expiry date on the label

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500


Still not finding the right product?

Explore all of our products under Gram′s Iodine Solution


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters