Skip to Content

HT10132

Bouin′s solution

histological fixative

Synonym(s):

Bouin’s fluid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12171500
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shelf life

Expiry date on the label

composition

acetic acid, 5% , formaldehyde, 9% , picric acid, 0.9%

color

yellow

application(s)

hematology
histology

storage temp.

room temp

General description

Bouin′s solution is a noncoagulant yellow stain fixative that consists of saturated picric acid, formaldehyde, and acetic acid.

Application

Bouin′s solution is an excellent fixative for preserving soft and delicate structures. It is used as a mordant in various trichrome procedures.
Additionally, it has been used for the following purposes-
  • staining of human liver biopsy tissue samples for two-photon excited fluorescence (TPEF) and second-harmonic generation (SHG) imaging
  • fixation of rat ovaries for the observing follicle cells using microscopy
  • fixation of gonads of crab for morphological and histological analysis
Prepared with saturated picric acid, formaldehyde and acetic acid. Excellent fixative for preserving soft and delicate structures. Used as a mordant in various trichrome procedures.

Biochem/physiol Actions

Bouin′s solution causes shrinkage of tissues when used as a preservative due to its picric acid component. It preserves morphological and nuclear information in tissues upon fixation.


Still not finding the right product?

Explore all of our products under Bouin′s solution


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library