HT1004
Gold Chloride Solution
200 mg/dL in deionized water
Synonym(s):
Gold chloride solution
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
solution
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
concentration
200 mg/dL in deionized water
application(s)
hematology
histology
storage temp.
2-8°C
SMILES string
Cl.Cl[Au](Cl)Cl
InChI
1S/Au.4ClH/h;4*1H/q+3;;;;/p-3
InChI key
VDLSFRRYNGEBEJ-UHFFFAOYSA-K
Application
Used in procedure HT100 for the toning of sections stained with the Silver Methanamine procedure
Components
Gold chloride, 0.2% in aqueous solution
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nature communications, 12(1), 639-639 (2021-01-29)
The generation of toxic amyloid β (Aβ) oligomers is a central feature of the onset and progression of Alzheimer's disease (AD). Drug discoveries for Aβ oligomer degradation have been hampered by the difficulty of Aβ oligomer purification and a lack
Proceedings of the National Academy of Sciences of the United States of America, 115(41), E9717-E9726 (2018-09-23)
Surface protein dynamics dictate synaptic connectivity and function in neuronal circuits. ASTN2, a gene disrupted by copy number variations (CNVs) in neurodevelopmental disorders, including autism spectrum, was previously shown to regulate the surface expression of ASTN1 in glial-guided neuronal migration.
eLife, 10 (2021-02-16)
Cholinergic fast time-scale modulation of cortical physiology is critical for cognition, but direct local measurement of neuromodulators in vivo is challenging. Choline oxidase (ChOx)-based electrochemical biosensors have been used to capture fast cholinergic signals in behaving animals. However, these transients
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service