Merck
HS15994F
Premium slide box
100 place, cork lining, gray plastic
Synonym(s):
Schiebekasten, slide box microscope, slide storage, slide storage box
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.17
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS15994F
Feature:
Holds 100 x slides 76x26 mm
Material:
gray plastic , nickel closure
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
gray plastic , nickel closure
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994F
L × W × H
208 × 175 × 34 mm
Features and Benefits
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
  • Molded, rigid boxes securely hold slides in grooved slots
  • Durable ABS plastic boxes are available with cork lining only
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Feature thumb-latch lock mechanism
Still not finding the right product?
Explore all of our products under Premium slide box
Choose from one of the most recent versions:
Certificates of Analysis (COA)