Merck
HS15906
Spatula with Flat End & Spoon End
stainless steel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.71
UNSPSC Code:
41122403
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
stainless steel
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS15906
L × W
150 mm × 3.8 mm
General description
Spatula transfers, lifts, and handles powders and granular materials.
Features and Benefits
  • Made from nickel-plated, stainless steel wire
  • Glazed finish with a Hayman-type end (flat and spoon)
  • Autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)