Skip to Content
HS100503
Gusto® High Speed Mini Centrifuge Rotor
to hold, 12 x 1.5/2 mL tubes
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.78
UNSPSC Code:
41103909
Manufacturer/tradename:
Heathrow Scientific HS100503
Parameter:
9,800 × g max. RCF
Feature:
to hold 12 x 1.5/2 mL tubes
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
to hold 12 x 1.5/2 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific HS100503
parameter
9,800 × g max. RCF
General description
Additional 12 place 1.5/2.0 mL tube rotor for Gusto High Speed Mini Centrifuge
Packaging
INCLUDES:
Cover and Knob
Legal Information
GUSTO is a registered trademark of Heathrow Scientific
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)