Skip to Content
Merck

HS1002A

Autoclave disposal bags

H × W × thickness 660 mm × 310 mm × 2.0 mm, transparent polypropylene (with blue biohazard labeling )

Synonym(s):

autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.17
UNSPSC Code:
41106621
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

transparent polypropylene (with blue biohazard labeling )

feature

label blue biohazard labeling

manufacturer/tradename

Heathrow Scientific HS1002A

packaging

pkg of -200 bags (4 packs of 50 bags)

H × W × thickness

660 mm × 310 mm × 2.0 mm

General description

Autoclave Bags are clear, imprinted with blue biohazard labeling designed for safe disposal.
Constructed of 2.0 mm thick polypropylene, allowing for repeated autoclaving
  • Strong side seams for securely holding items
  • Front write-on panel
  • Autoclavable at 250°F (121°C).
  • 4 inner packs of 50 bags, Sold as a case of 200
  • Disposable

Autoclave disposal bags are clear, imprinted with blue biohazard labeling designed for safe disposal. It is made of 2.0 mm thick polypropylene and can be autoclaved multiple times.

Features and Benefits

  • Strong side seams for securing objects
  • Front write-on panel
  • Autoclavable at 250°F (121°C).
  • 4 inner packs of 50 bags in a case of 200


Still not finding the right product?

Explore all of our products under Autoclave disposal bags




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library