Skip to Content
Merck
All Photos(7)

Documents

HPA043420

Sigma-Aldrich

Anti-UMOD antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ADMCKD2, Anti-ADTKD1, Anti-FJHN, Anti-HNFJ, Anti-HNFJ1, Anti-MCKD2, Anti-THGP, Anti-THP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GYYVYNLTAPPECHLAYCTDPSSVEGTCEECSIDEDCKSNNGRWHCQCKQDFNITDISLLEHRLECGANDMKVSLGK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UMOD(7369)

General description

Uromodulin (UMOD) is a glycoprotein produced particularly in renal tubular cells and released into urine. The gene is located on human chromosome 16p12.3. It is the most abundant protein in urine.

Immunogen

uromodulin

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Uromodulin (UMOD) plays an important role in inflammatory responses and acts as a potential therapeutic for hypertension. It provides innate immunity and protects from kidney stones. The protein provides protection from uropathogenic bacteria. Mutations in this gene is associated with inflammation and development of kidney diseases, such as medullary cystic kidney disease 2 (MCKD2), glomerulocystic kidney disease with hyperuricemia and isosthenuria (GCKDHI) and familial juvenile hyperuricemic nephropathy (FJHN).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82316

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional analysis of UMOD gene and its effect on inflammatory cytokines in serum of essential hypertension patients
Jian L, et al.
International Journal of Clinical and Experimental Pathology, 8(9), 11356-11356 (2015)
Modulation of urinary peptidome in humans exposed to high altitude hypoxia
Mainini V, et al.
Molecular Biosystems, 8(4), 959-966 (2012)
Uromodulin rs4293393 T> C variation is associated with kidney disease in patients with type 2 diabetes
Kumar V, et al.
The Indian Journal of Medical Research, 146(2), S15-S15 (2017)
Unravelling the genetic basis of renal diseases; from single gene to multifactorial disorders
Kumar V, et al.
The Journal of Pathology, 146(Suppl 2), S15-S15 (2017)
A structured interdomain linker directs self-polymerization of human uromodulin
Bokhove M, et al.
Proceedings of the National Academy of Sciences of the USA, 113(6), 1552-1557 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service