Skip to Content
Merck
All Photos(6)

Key Documents

HPA031752

Sigma-Aldrich

Anti-WRNIP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ22526, Anti-WHIP, Anti-Werner helicase interacting protein 1, Anti-bA420G6.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

YQGCHFIGMPECEVLLAQCVVYFARAPKSIEVYSAYNNVKARLRNHQGPLPPVPLHLRNAPTRLMKDLGYG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WRNIP1(56897)

General description

The gene WRNIP1 (werner helicase-interacting protein 1) is mapped to human chromosome 6p25.2. The encoded protein is from AAA+ class of ATPase family proteins. It is an evolutionarily conserved protein. The protein has a small Rad18 (radiation-sensitive 18)-like zinc finger at the amino terminus and a AAA+ ATPase domain in the center of the protein.

Immunogen

Werner helicase interacting protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

WRNIP1 (werner helicase-interacting protein 1) might be involved in the stimulation of DNA polymerase δ to restart DNA synthesis during fork arrest. Absence of the gene results in the accumulation of DNA damages. WRNIP1 helps in providing protection to stalled forks from degradation, partly by stabilizing RAD51 (DNA repair protein) on single stranded DNA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77437

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner helicase-interacting protein 1 binds polyubiquitin via its zinc finger domain.
Bish RA
The Journal of Biological Chemistry, 282, 23184-23193 (2007)
WRNIP1 protects stalled forks from degradation and promotes fork restart after replication stress.
Leuzzi G
The Embo Journal, 35, 1437-1451 (2016)
Molecular genetics of adult ADHD: converging evidence from genome-wide association and extended pedigree linkage studies.
Lesch KP
Journal of neural transmission (Vienna, Austria : 1996), 115, 1573-1573 (2008)
Åsa Ehlén et al.
Translational oncology, 4(4), 212-221 (2011-08-02)
The RNA-binding motif protein 3 (RBM3) was initially discovered as a putative cancer biomarker based on its differential expression in various cancer forms in the Human Protein Atlas (HPA). We previously reported an association between high expression of RBM3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service