Skip to Content
Merck
All Photos(7)

Key Documents

HPA030782

Sigma-Aldrich

Anti-MKL1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BSAC, Anti-KIAA1438, Anti-MAL, Anti-MRTF-A, Anti-megakaryoblastic leukemia (translocation) 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EQEKRAQQPAPAPAPLGTPVKQENSFSSCQLSQQPLGPAHPFNPSLAAPATNHIDPCAVAPGPPSVVVKQEALQPEPEPVPAPQLLLGPQGPSLIKGVAPPTLITDSTGTHLVLTVTNKNADSPG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MKL1(57591)

General description

The gene MKL1 (megakaryoblastic leukemia (translocation) 1) is mapped to human chromosome 22q13.2. It belongs to the myocardin-related family of transcriptional coactivators. The protein is present in the cytoplasm and nucleus. It is widely expressed in human tissues.

Immunogen

megakaryoblastic leukemia (translocation) 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-MKL1 antibody produced in rabbit has been used in immunofluorescence.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

MKL1 (megakaryoblastic leukemia (translocation) 1) is a transcriptional coactivator. G-actin interacts with MKL1 and prevents its association with transcription factor SRF (serum response factor). Release of MKL1 from G-actin results in an interaction between SRF and MKL1. This association controls the expression of cytoskeletal proteins. MKL1 is also involved in skeletal, cardiac and smooth muscle differentiation. It also enhances tumor cell invasion and metastasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73615

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsiao-Yen Ma et al.
The European respiratory journal (2022-12-31)
Idiopathic pulmonary fibrosis (IPF) is a progressive fibrotic disease characterized by aberrant fibroblast/myofibroblast accumulation and excessive collagen matrix deposition in the alveolar areas of lungs. As the first approved IPF medication, pirfenidone (PFD) significantly decelerates lung function decline while its
Stefania Petrini et al.
Aging, 9(5), 1453-1469 (2017-06-01)
Nuclear integrity and mechanical stability of the nuclear envelope (NE) are conferred by the nuclear lamina, a meshwork of intermediate filaments composed of A- and B-type lamins, supporting the inner nuclear membrane and playing a pivotal role in chromatin organization
Qiande Hu et al.
Cell & bioscience, 11(1), 25-25 (2021-01-30)
Alveolar Rhabdomyosarcoma (ARMS) is a pediatric malignant soft tissue tumor with skeletal muscle phenotype. Little work about skeletal muscle proteins in ARMS was reported. PAX3-FOXO1 is a specific fusion gene generated from the chromosomal translocation t (2;13) (q35; q14) in
Adriana Sánchez-Danés et al.
Nature, 562(7727), 434-438 (2018-10-10)
Basal cell carcinoma (BCC) is the most frequent cancer in humans and results from constitutive activation of the Hedgehog pathway1. Several Smoothened inhibitors are used to treat Hedgehog-mediated malignancies, including BCC and medulloblastoma2. Vismodegib, a Smoothened inhibitor, leads to BCC
Michaela-Rosemarie Hermann et al.
Journal of cell science, 129(7), 1391-1403 (2016-02-14)
Integrin-mediated activation of small GTPases induces the polymerisation of G-actin into various actin structures and the release of the transcriptional co-activator MRTF from G-actin. Here we report that pan-integrin-null fibroblasts seeded on fibronectin and expressing β1- and/or αV-class integrin contained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service