Skip to Content
Merck
All Photos(5)

Key Documents

HPA029874

Sigma-Aldrich

Anti-CAMP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAP18, Anti-FALL-39, Anti-FALL39, Anti-LL37, Anti-cathelicidin antimicrobial peptide

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

RSSDANLYRLLDLDPRPTMDGDPDTPKPVSFTVKETVCPRTTQQSPEDCDFKKDGLVKRCMGTVTLNQARGSFDISCDKDNKRFALLGDFFRKSKEKIGKEFKRIVQRIKDFLRNLVPR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMP(820)

General description

CAMP (cationic antimicrobial peptide) is a necessary component of human innate immunity. It is also known as LL-37 (antimicrobial peptide). It is produced by various cells, including epithelial cells, keratinocytes, and neutrophils. CAMPs are designated by α- and β-defensins and the cathelicidin-derived LL-37 peptide. It is mostly present in microenvironments, that foster carbamylation. It is located on chromosome 3p21.

Immunogen

cathelicidin antimicrobial peptide recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CAMP has been used in IF (immunofluorescence) and in immunohistochemistry.

Biochem/physiol Actions

CAMP (cationic antimicrobial peptides) kills a wide spectrum of pathogenic bacteria. It neutralizes LPS (lipopolysaccharide), and act as a chemoattractant for immune cells including T cells, monocytes, neutrophils, and mast cells. It enormously affects the direction of dendritic cell maturation, activates production of cytokines, chemokines, and their receptors and stimulates mast cell degranulation. Its major role is to act as a signaling molecule for innate and acquired immunity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72145

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphism in cathelicidin gene (CAMP) that alters Hypoxia-inducible factor (HIF-1?::ARNT) binding is not associated with tuberculosis
Lopez Campos GN
International Journal of Immunogenetics, 41(1), 54-62 (2014)
Carbamylated LL-37 as a modulator of the immune response
Koro C, et al.
Innate Immunity, 22(3), 218-229 (2016)
Tiziana Cubeddu et al.
Veterinary immunology and immunopathology, 189, 66-70 (2017-07-04)
Cathelicidins are well-characterized antimicrobial peptides (AMPs) that are present in significant amounts in mastitic milk. Neutrophils are believed to be the main producers of these AMPs, while the role of mammary epithelial cells (MECs) in their production and release is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service