Skip to Content
Merck
All Photos(7)

Key Documents

HPA023313

Sigma-Aldrich

Anti-GSDMA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Gasdermin-1, Anti-Gasdermin-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

VGALTELSEAQQKLLVKSMEKKILPVQLKLVESTMEQNFLLDKEGVFPLQPELLSSLGDEELTLTEALVGLSGLEVQRSGPQYMWDPDTLPRLCALYAGLSLLQQLTKAS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSDMA(284110)

General description

The gene GSDMA (gasdermin A) is mapped to human chromosome 17q21. The protein localizes in the cytoplasm.

Immunogen

Gasdermin-A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GSDMA (gasdermin A) is associated with childhood asthma. It is present in the upper gastrointestinal tract and controls apoptosis of the gastric epithelium. GSDMA is downregulated in gastric cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75781

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David R Crosslin et al.
Human genetics, 131(4), 639-652 (2011-11-01)
White blood cell count (WBC) is unique among identified inflammatory predictors of chronic disease in that it is routinely measured in asymptomatic patients in the course of routine patient care. We led a genome-wide association analysis to identify variants associated
N Saeki et al.
Oncogene, 26(45), 6488-6498 (2007-05-02)
Defining apoptosis-regulatory cascades of the epithelium is important for understanding carcinogenesis, since cancer cells are considered to arise as a result of the collapse of the cascades. We previously reported that a novel gene GASDERMIN (GSDM) is expressed in the
Jinho Yu et al.
Pediatric pulmonology, 46(7), 701-708 (2011-02-22)
The gasdermin A (GSDMA) and gasdermin B (GSDMB) genes are located at 17q21.2. The GSDM family genes have been studied in the gastrointestinal tract but recent reports suggest that GSDMB is associated with childhood asthma in several populations. We investigated
Julia Lachner et al.
Scientific reports, 7(1), 17446-17446 (2017-12-14)
The homeostasis of the epidermis depends on keratinocyte differentiation and cornification, a mode of programmed cell death that does not elicit inflammation. Here, we report that cornification is associated with the expression of specific genes that control multiple steps of
Norihisa Saeki et al.
Genes, chromosomes & cancer, 48(3), 261-271 (2008-12-04)
Gasdermin (GSDM or GSDMA), expressed in the upper gastrointestinal tract but frequently silenced in gastric cancers (GCs), regulates apoptosis of the gastric epithelium. It has three human homologs, GSDMB, GSDMC, and GSDMD (GSDM family) and they are considered to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service