Skip to Content
Merck
All Photos(8)

Key Documents

HPA020108

Sigma-Aldrich

Anti-PAEP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GD, Anti-Glycodelin, Anti-PAEG, Anti-PEG, Anti-PP14, Anti-Placental protein 14, Anti-Pregnancy-associated endometrial alpha-2 globulin, Anti-Progesterone-associated endometrial protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

NYTVANEATLLDTDYDNFLFLCLQDTTTPIQSMMCQYLARVLVEDDEIMQGFIRAFRPLPRHLWYLLDLKQME

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAEP(5047)

General description

PAEP is a glycoprotein that is synthesized in the secretory and decidualized endometrium in women. This protein is also secreted by the seminal vesicle epithelium in men. PAEP gene is located on the human chromosome at 9q34.3.
Progesterone-associated endometrial protein (PAEP) is a glycoprotein which belongs to the lipocalin family. It exists as four isoforms and possesses a characteristic lipocalin fold.

Immunogen

Glycodelin Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Downregulation of the PAEP gene is associated with increased activation of the maternal immune system which can lead to abortion during the first trimester. Overexpression of the PAEP gene is seen in ovarian and breast cancer. PAEP is also observed in lung cancer and melanoma cells.
Progesterone-associated endometrial protein (PAEP) acts as an immunosuppressant. It takes part in reproduction and is expressed in endometrial cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75226

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tine Cuppens et al.
International journal of cancer, 142(6), 1230-1243 (2017-10-25)
Uterine leiomyosarcomas (uLMS) are rare, aggressive malignancies for which limited treatment options are available. To gain novel molecular insights into uLMS and identify potential novel therapeutic targets, we characterized 84 uLMS samples for genome-wide somatic copy number alterations, mutations, gene
M Kämäräinen et al.
Blood, 84(2), 467-473 (1994-07-15)
Progesterone-associated endometrial protein (PAEP/PP14) is a 28-kD glycoprotein with sequence homology to beta-lactoglobulins containing a retinol-binding motif. PAEP/PP14 is present at nanomolar concentrations in human serum. It is produced by secretory and decidualized endometrium in women and by seminal vesicle
Marc A Schneider et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(15), 3529-3540 (2015-04-23)
In recent years, immune therapeutic strategies against non-small cell lung cancer (NSCLC) based on tissue-derived biomarkers, for example PD1/PD-L1 (CD274), have evolved as novel and promising treatment options. However, the crosstalk between tumor and immune cells is poorly understood. Glycodelin
André Schiefner et al.
The Biochemical journal, 466(1), 95-104 (2014-11-26)
Human glycodelin (Gd) is an abundant glycoprotein from the lipocalin family and is involved in crucial biological processes such as reproduction and immune reaction. In females and males, Gd is found in four distinct glycoforms-A, C, F and S-that arise
Miriam Lenhard et al.
BMC cancer, 13, 616-616 (2014-01-01)
Knowledge on immunosuppressive factors in the pathogenesis of endometrial cancer is scarce. The aim of this study was to assess Glycodelin (Gd) and its immunosuppressive isoform Glycodelin A (GdA) in endometrial cancer tissue and to analyze its impact on clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service