Skip to Content
Merck
All Photos(8)

Key Documents

HPA018991

Sigma-Aldrich

Anti-TUFM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EF-Tu, Anti-Elongation factor Tu, mitochondrial precursor, Anti-P43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LSKEEGGRHKPFVSHFMPVMFSLTWDMACRIILPPEKELAMPGEDLKFNLILRQPMILEKGQRFTLRDGNRTIGTGLVTNTLAMTEEEKNIKWG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUFM(7284)

General description

The gene TUFM (Elongation factor Tu - mitochondrial) is mapped to human chromosome 16p11.2. The protein localizes in the mitochondria.

Immunogen

Elongation factor Tu, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-TUFM antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. In addition, the antibody has been used for co-immunoprecipitation and immunocytochemistry.

Biochem/physiol Actions

TUFM (Elongation factor Tu - mitochondrial) is a GTPase. It forms a ternary complex with GTP (Guanosine-5′-triphosphate) and aminoacylated (aa)-tRNA and causes binding of the aa-tRNA to the A-site of the ribosome. Mutations in TUFM cause infantile encephalopathy and defective mitochondrial DNA translation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74747

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Lei et al.
Immunity, 36(6), 933-946 (2012-07-04)
The mitochondrial protein MAVS (also known as IPS-1, VISA, and CARDIF) interacts with RIG-I-like receptors (RLRs) to induce type I interferon (IFN-I). NLRX1 is a mitochondrial nucleotide-binding, leucine-rich repeats (NLR)-containing protein that attenuates MAVS-RLR signaling. Using Nlrx1(-/-) cells, we confirmed
Binbin Ding et al.
Cell host & microbe, 21(4), 538-547 (2017-04-14)
Mitophagy is a form of autophagy that selectively removes damaged mitochondria. Impaired mitochondria can be tagged by the kinase PINK1, which triggers recruitment of the E3-ubiquitin ligase Parkin and subsequent mitochondrial sequestration within autophagosomes. We previously found that human parainfluenza
Rikke Raaen Lund et al.
Proteomics, 12(13), 2139-2148 (2012-05-25)
The development of metastasis is a complex, multistep process that remains poorly defined. To identify proteins involved in the colonization phase of the metastatic process, we compared the proteome of tumors derived from inoculation of a panel of isogenic human
Lucia Valente et al.
American journal of human genetics, 80(1), 44-58 (2006-12-13)
Mitochondrial protein translation is a complex process performed within mitochondria by an apparatus composed of mitochondrial DNA (mtDNA)-encoded RNAs and nuclear DNA-encoded proteins. Although the latter by far outnumber the former, the vast majority of mitochondrial translation defects in humans
Lucia Valente et al.
Biochimica et biophysica acta, 1792(8), 791-795 (2009-06-16)
The mitochondrial translational machinery allows the genes encoded by mitochondrial DNA (mtDNA) to be translated in situ. Mitochondrial translation requires a number of nucleus-encoded protein factors, some of which have been found to carry mutations in patients affected by mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service