Skip to Content
Merck
All Photos(7)

Key Documents

HPA015650

Sigma-Aldrich

Anti-RTN3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NSP-like protein II, Anti-NSPLII, Anti-Neuroendocrine-specific protein-like 2, Anti-Reticulon-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

IMTSSFLSSSEIHNTGLTILHGEKSHVLGSQPILAKEGKDHLDLLDMKKMEKPQGTSNNVSDSSVSLAAGVHCDRPSIPASFPEHPAFLSKKIGQVEEQIDKETKNPNGVSSREAKTALDADDRFTLLTAQKPPTEY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RTN3(10313)

Immunogen

Reticulon-3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Reticulon-3 is a protein encoded by the RTN3 gene in humans and belongs to reticulon family. This protein is found to interact with BACE1 (β-site amyloid precursor protein cleaving enzyme 1) and its Aβ-generating activity is inhibited. It may act as a candidate protein in regulating the pathogenesis of atherosclerosis (AS) and Alzheimer′s disease. It may also be involved in axonal transport (increase of this protein causes its imbalance) and its accumulation in swollen neuritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72724

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wataru Araki et al.
Current Alzheimer research, 10(2), 135-142 (2012-06-30)
Inhibition of the β-secretase, BACE1, which cleaves amyloid precursor protein (APP) to produce β-amyloid protein (Aβ), is thought to be a feasible therapeutic strategy for Alzheimer's disease. Reticulon (RTN) proteins such as RTN3 have been identified as membrane proteins that
Yaqin Chen et al.
Molecular and cellular biochemistry, 361(1-2), 67-70 (2011-10-04)
The recruitment of monocytes to arterial wall and their transformation into macrophages are generally accepted as important early events in the pathogenesis of atherosclerosis (AS). Our research group found Reticulon3 (RTN3), a member of the reticulon family, may be a
Qi Shi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(16), 5108-5115 (2009-04-24)
Reticulon 3 (RTN3) has been shown to mark a distinct and abundant population of dystrophic neurites named RTN3 immunoreactive dystrophic neurites (RIDNs) in patients' brains of Alzheimer disease (AD). Transgenic mice expressing RTN3 (Tg-RTN3) also spontaneously develop RIDNs. To determine
Haoxi Wu et al.
Developmental cell, 56(1), 52-66 (2021-01-13)
ER tubules form and maintain membrane contact sites (MCSs) with endosomes. How and why these ER-endosome MCSs persist as endosomes traffic and mature is poorly understood. Here we find that a member of the reticulon protein family, Reticulon-3L (Rtn3L), enriches
Qi Shi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(7), 3059-3066 (2013-02-15)
Neuritic dystrophy is one of the important pathological features associated with amyloid plaques in Alzheimer's disease (AD) and age-dependent neuronal dysfunctions. We reported previously that reticulon-3 (RTN3) immunoreactive dystrophic neurites (RIDNs) are abundantly present in the hippocampus of AD patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service