Skip to Content
Merck
All Photos(2)

Key Documents

HPA014488

Sigma-Aldrich

Anti-CCR6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-C CKR-6, Anti-C-C chemokine receptor type 6, Anti-CC-CKR-6, Anti-CCR-6, Anti-CD196 antigen, Anti-CKR-L3, Anti-Chemokine receptor-like 3, Anti-DRY6, Anti-G-protein coupled receptor 29, Anti-GPR-CY4, Anti-GPRCY4, Anti-LARC receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

KYNTQGSDVCEPKYQTVSEPIRWKLLML

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCR6(1235)

General description

The gene CCR6 (C-C motif chemokine receptor 6) encodes a chemokine receptor that is mainly expressed in B cells and memory T cells. It is also found to be expressed in dendritic cells, spleen, thymus, small intestine, and peripheral blood leukocytes. The gene is mapped to human chromosome 6q27. It shares sequence similarity with G protein-coupled chemokine receptor. The acidic amino terminal contains two potential N-linked glycosylation sites, and two additional ones are located in the first and the third extracellular loops.

Immunogen

C-C chemokine receptor type 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene CCR6 (C-C motif chemokine receptor 6) encodes a chemokine receptor for the chemokine ligand CCL20, also referred to as LARC (liver and activation-regulated chemokine) or MIP-3 α (macrophage inflammatory protein-3 α). The binding of the ligand to the receptor facilitates the chemoattraction of immature dendritic cells, effector/memory T-cells and B-cells. It participates in pathology, including cancer and rheumatoid arthritis. In mucosal tissue, CCR6 participates in the regulation of humoral immunity and lymphocyte homeostasis. It also mediates dendritic cell localization.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72741

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malte A Kluger et al.
Journal of the American Society of Nephrology : JASN, 25(6), 1291-1302 (2014-02-11)
A pathogenic role for Th17 cells in inflammatory renal disease is well established. The mechanisms underlying their counter-regulation are, however, largely unknown. Recently, Th17 lineage-specific regulatory T cells (Treg17) that depend on activation of the transcription factor Stat3 were identified.
D N Cook et al.
Immunity, 12(5), 495-503 (2000-06-08)
Chemokine-directed migration of leukocyte subsets may contribute to the qualitative differences between systemic and mucosal immunity. Here, we demonstrate that in mice lacking the chemokine receptor CCR6, dendritic cells expressing CD11c and CD11b are absent from the subepithelial dome of
Evemie Schutyser et al.
Cytokine & growth factor reviews, 14(5), 409-426 (2003-09-02)
CCL20, alternatively named liver and activation-regulated chemokine (LARC), macrophage inflammatory protein-3alpha (MIP-3alpha) or Exodus-1, is the only chemokine known to interact with CC chemokine receptor 6 (CCR6), a property shared with the antimicrobial beta-defensins. The ligand-receptor pair CCL20-CCR6 is responsible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service