Skip to Content
Merck
All Photos(3)

Key Documents

HPA014101

Sigma-Aldrich

Anti-CHRM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Muscarinic acetylcholine receptor M1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

SETPGKGGGSSSSSERSQPGAEGSPETPPGRCCRCCRAPRLLQAYSWKEEEEEDEGSMESLTSSEGEEPGSEVVIKMPMVDPEAQAPTKQPPRSSPNTVKRPTKKGRDRAGKGQKPRGKEQLAKRKTFSLVKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRM1(1128)

General description

The gene CHRM1 (cholinergic receptor, muscarinic 1) encodes a muscarinic cholinergic receptor that is the most abundant muscarinic receptor in the CNS (central nervous system). It is predominantly expressed in the neurons. It is a member of the muscarinic acetylcholine receptor family that conatins five subtypes, CHRM1-CHRM5. These subtypes have high homology but differ in signal transduction mediated via G protein-coupled pathways, resulting in varying physiological responses. The subtypes CHRM1, CHRM3, and CHRM5 have affinity for G proteins of the Gq/11 family and CHRM2 and CHRM4 bind to Gi/o-type G proteins. The gene is mapped to human chromosome 11q13.

Immunogen

Muscarinic acetylcholine receptor M1 recombinant protein epitope signature tag (PrEST)

Application

Anti-CHRM1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

The protein encoded by the gene CHRM1 (cholinergic receptor, muscarinic 1) stimulates intracellular Ca2+ signaling in response to acetylcholine in striatal cells. The expression of this protein is modulated by heterochromatin condensation and any changes in its structure leads to impaired Ca2+ release from ER. The encoded protein enhances phosphatidylinositol (PtdIns) hydrolysis and mobilization of intracellular Ca2+. Both CHRM1 and CHRM2 are involved in regulating cholinergic action on glutamatergic and GABAergic nerve terminals in the stiratum and this system has been associated with HD (Huntington′s disease) pathology.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72768

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C N Adra et al.
Clinical genetics, 55(6), 431-437 (1999-08-18)
Asthma is a complex syndrome in which bronchial inflammation and smooth muscle hyperactivity lead to labile airflow obstruction. The commonest form of asthma is that due to atopy, which is an immune disorder where production of IgE to inhaled antigens
Junghee Lee et al.
Acta neuropathologica, 125(5), 727-739 (2013-03-05)
Huntington's disease (HD) is an autosomal dominant neurodegenerative disease caused by an expanded trinucleotide CAG repeat in the gene coding for huntingtin. Deregulation of chromatin remodeling is linked to the pathogenesis of HD but the mechanism remains elusive. To identify
Judith Molina et al.
Diabetes, 63(8), 2714-2726 (2014-03-25)
Acetylcholine regulates hormone secretion from the pancreatic islet and is thus crucial for glucose homeostasis. Little is known, however, about acetylcholine (cholinergic) signaling in the human islet. We recently reported that in the human islet, acetylcholine is primarily a paracrine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service