Skip to Content
Merck
All Photos(1)

Documents

HPA013955

Sigma-Aldrich

Anti-GPR19 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GPR-NGA, Anti-Probable G-protein coupled receptor 19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RMDNSKPHLIIPTLLVPLQNRSCTETATPLPSQYLMELSEEHSWMSNQTD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR19(2842)

General description

The gene GPR19 (G protein-coupled receptor 19) encodes a receptor having sequence similarity with the D2 dopamine and neuropeptide Y families of receptors. The gene is mapped to human chromosome 12. It is found to be expressed in peripheral and brain regions similar to the D2 receptor gene expression pattern.

Immunogen

Probable G-protein coupled receptor 19 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene GPR19 (G protein-coupled receptor 19) is localized to a region on the human chromosome that is commonly rearranged in cancers, such as hematological malignancies and some solid neoplasias. This region is also observed to be deleted in childhood acute lymphoblastic leukemia. The encoded protein belongs to the family of G-protein coupled receptors that contain the characteristic seven transmembrane domains. The protein may be involved in the early development of nervous system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72788

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B F O'Dowd et al.
FEBS letters, 394(3), 325-329 (1996-10-07)
Following the cloning of the dopamine receptors we continued a search of the human genome for related genes. We searched an EST data base and discovered cDNA fragments encoding novel G protein-coupled receptor genes. The available GenBank sequence of one
A Montpetit et al.
Human genetics, 105(1-2), 162-164 (1999-09-10)
Rearrangements of the short arm of chromosome 12 are frequently observed in hematological malignancies and in certain solid neoplasias. Loss of heterozygosity studies identified a small genetic interval on chromosome 12p12.3 that is frequently deleted in childhood acute lymphoblastic leukemia
Sabine A H Hoffmeister-Ullerich et al.
Cell and tissue research, 318(2), 459-463 (2004-09-29)
G-protein-coupled receptors (GPCRs) are characterized by seven transmembrane domains and constitute the largest and structurally best-conserved family of signaling molecules. They are present in a diversity of organs and tissues and are involved in virtually all physiological processes. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service