Skip to Content
Merck
All Photos(4)

Key Documents

HPA012582

Sigma-Aldrich

Anti-AZGP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc-alpha-2-glycoprotein precursor, Anti-Zn- alpha-2-GP, Anti-Zn-alpha-2-glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

QDGRYSLTYIYTGLSKHVEDVPAFQALGSLNDLQFFRYNSKDRKSQPMGLWRQVEGMEDWKQDSQLQKAREDIFMETLKDIVEYYNDSNGSHVLQGRFGCEIENNRSSGAFWKYYYDGKDYI

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AZGP1(563)

Immunogen

Zinc-alpha-2-glycoprotein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-AZGP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

AZGP1 (α-2-glycoprotein 1, zinc-binding) is a soluble, truncated secretory major histocompatibility complex-related protein involved in lipid degradation. It has structural homology with the class I major histocompatibility complex (MHC) heavy chain. It possesses molecular weight of 38,500 Mr. It consists of a large groove analogous to class I MHC peptide binding grooves. It has been suggested that AZGP1 may involve in the expression of the immune response.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84350

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mads Dochedahl Winther et al.
Diagnostics (Basel, Switzerland), 10(8) (2020-07-31)
Biomarkers for predicting the risk of castration-resistant prostate cancer (CRPC) in men treated with primary androgen deprivation therapy (ADT) are lacking. We investigated whether Zinc-alpha 2 glycoprotein (AZGP1) expression in the diagnostic biopsies of men with hormone-naïve prostate cancer (PCa)
James D Brooks et al.
The Prostate, 76(15), 1409-1419 (2016-06-22)
Given the uncertainties inherent in clinical measures of prostate cancer aggressiveness, clinically validated tissue biomarkers are needed. We tested whether Alpha-2-Glycoprotein 1, Zinc-Binding (AZGP1) protein levels, measured by immunohistochemistry, and RNA expression, by RNA in situ hybridization (RISH), predict recurrence
L M Sánchez et al.
Science (New York, N.Y.), 283(5409), 1914-1919 (1999-04-17)
Zn-alpha2-glycoprotein (ZAG) is a soluble protein that is present in serum and other body fluids. ZAG stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. The 2.8 angstrom crystal structure of ZAG resembles
Juan F García-Moreno et al.
Cellular and molecular life sciences : CMLS, 80(7), 185-185 (2023-06-21)
DIS3L2 degrades different types of RNAs in an exosome-independent manner including mRNAs and several types of non-coding RNAs. DIS3L2-mediated degradation is preceded by the addition of nontemplated uridines at the 3'end of its targets by the terminal uridylyl transferases 4
T Araki et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(3), 679-683 (1988-02-01)
In the present study the complete amino acid sequence of human plasma Zn-alpha 2-glycoprotein was determined. This protein whose biological function is unknown consists of a single polypeptide chain of 276 amino acid residues including 8 tryptophan residues and has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service