Skip to Content
Merck
All Photos(5)

Key Documents

HPA010928

Sigma-Aldrich

Anti-SFTPC Antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, rabbit polyclonal

Synonym(s):

Anti-BRICD6, Anti-PSP-C, Anti-SFTP2, Anti-SMDP2, Anti-SP-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

Product Name

Anti-SFTPC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

PEAQQRLALSEHLVTTATFSIGSTGLVVYDYQQLLIAYKPAPGTCCYIMKIAPESIPSLEALTRKVHNFQAKPAVPTSKLGQAEGRDAGSAPSGGDPAFLGMAVSTLCG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFTPC(6440)

General description

Pulmonary surfactant is a mixture of phospholipids and protein, which is surface-active and secreted by type II epithelial cells into the alveolar space. There are four proteins present in the surfactant namely, SP (surfactant protein)-A, SP-B, SP-C or SFTPC and SP-D. SFTPC is a hydrophobic protein, which is produced as a proprotein and undergoes post-translational modification to have its NH2 and COOH propeptides removed. The mature active protein has a molecular weight of 3.7kDa. The proprotein is composed of 197 amino acids and is produced exclusively by alveolar type 2 (AT2) cells. SFTPC contains BRICHOS domain, which is suggested to prevent the aggregation of the peptide and regulate proteolytic processing of SFTPC proprotein. This gene is located on human chromosome 8p21.

Immunogen

Pulmonary surfactant-associated protein C Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SFTPC antibody produced in rabbit has been used in immunohistochemistry (IHC) and immunofluorescence (IF).

Biochem/physiol Actions

SFTPC (surfactant protein C) stabilizes the pulmonary surfactant film and aids in the adsorption of phospholipids on the surfactant monolayer. This way it regulates the metabolism and dynamics of the phospholipids constituting pulmonary surfactant. Mutations in this gene are associated with interstitial lung disease (ILD) in infants. Mutations in SFTPC can lead to various disorders such as, chronic pneumonitis in infants, and usual or desquamative interstitial pneumonia and idiopathic pulmonary fibrosis in adults. G100S mutation in the BRICHOS domain of SFTPC proprotein, leads to increased endoplasmic reticulum (ER) stress and cell apoptosis. Therefore, G100S mutation in this gene is involved in the pathogenesis of familial pulmonary fibrosis in Japanese population. Mutations in SFTPC gene also result in combined pulmonary fibrosis and emphysema (CPFE) syndrome, which is characterized by emphysema, inflammation and fibrosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72206

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul A Stevens et al.
Pediatric research, 57(1), 89-98 (2004-11-24)
Human surfactant protein C (hSP-C(1-197)) is synthesized as a 197 amino acid proprotein and cleaved to a mature 3.7 kD form. Although interstitial lung disease in patients with mutations of the hSP-C gene is becoming increasingly recognized, the mechanisms linking
Recapitulating idiopathic pulmonary fibrosis related alveolar epithelial dysfunction in a human iPSC-derived air-liquid interface model
Schruf E, et al.
Faseb Journal, 103498-103498 (2020)
Variable phenotype associated with SP-C gene mutations: fatal case with the I73T mutation.
S Percopo et al.
The European respiratory journal, 24(6), 1072-1073 (2004-12-02)
Christoph H Mayr et al.
EMBO molecular medicine, 13(4), e12871-e12871 (2021-03-03)
The correspondence of cell state changes in diseased organs to peripheral protein signatures is currently unknown. Here, we generated and integrated single-cell transcriptomic and proteomic data from multiple large pulmonary fibrosis patient cohorts. Integration of 233,638 single-cell transcriptomes (n = 61) across
Recombinant BRICHOS chaperone domains delivered to mouse brain parenchyma by focused ultrasound and microbubbles are internalized by hippocampal and cortical neurons
Galan-Acosta L, et al.
Molecular and Cellular Neurosciences, 103498-103498 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service