Skip to Content
Merck
All Photos(1)

Key Documents

HPA010866

Sigma-Aldrich

Anti-BAMBI antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), Anti-NMA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

NKRLQDQRQQMLSRLHYSFHGHHSKKGQVAKLDLECMVPVSGHENCCLTCDKMRQADLSNDKILSLVHWGMYSGHGKLEFV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAMBI(25805)

General description

BAMBI (bone morphogenetic protein and activin membrane-bound inhibitor) is a pseudoreceptor of TGF-β, and is highly conserved in vertebrates. This protein is a transmembrane glycoprotein and shows homology with TGF-β type I receptors. It contains 260 amino acids. This gene is located on the human chromosome 10p12.3-p11.2.

Immunogen

BMP and activin membrane-bound inhibitor homolog (Xenopus laevis) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BAMBI (bone morphogenetic protein and activin membrane-bound inhibitor) acts as a target for β-catenin, and suppresses adipogenesis through Wnt/β-catenin signaling pathway. It is up-regulated in various malignancies such as liver, ovarian, thyroid and colorectal cancer. Its overexpression in gastric cancer is associated with tumor metastasis and progression, relapse and poor survival rates in patients. By regulating the expression of β-catenin, BAMBI also plays a role in the tumorigenesis and progression of osteosarcoma. It is highly expressed in the lungs of chronic obstructive pulmonary disease (COPD) patients. Nontypeable Haemophilus influenzae (NTHI) infection up-regulates BAMBI gene, which might lead to improper repair of lung tissue and profibrotic activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71703

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Marwitz et al.
Cancer research, 76(13), 3785-3801 (2016-05-20)
Non-small cell lung cancer (NSCLC) is characterized by early metastasis and has the highest mortality rate among all solid tumors, with the majority of patients diagnosed at an advanced stage where curative therapeutic options are lacking. In this study, we
Lu Zhou et al.
Oncology reports, 30(3), 1315-1322 (2013-06-29)
The pseudoreceptor BAMBI (bone morphogenetic protein and activin membrane-bound inhibitor), formerly known as NMA, is an inhibitor of the TGF-β signaling pathway. BAMBI exhibits structural homology to TGF-βRI but lacks an intracellular kinase domain. In most of the high-grade carcinomas
Yin Mai et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 92(3), 172-182 (2014-05-07)
The process of differentiation from preadipocytes to adipocytes contributes to adipose tissue expansion in obesity. Blocking adipogenesis may be conducive to the etiology of obesity-related diseases. BMP and activin membrane-bound inhibitor (BAMBI) is a transmembrane protein, which was identified as
Daniel Drömann et al.
Respiratory research, 11, 67-67 (2010-06-02)
Nontypeable Haemophilus influenzae (NTHI) may play a role as an infectious trigger in the pathogenesis of chronic obstructive pulmonary disease (COPD). Few data are available regarding the influence of acute and persistent infection on tissue remodelling and repair factors such
Hyo-Seol Lee et al.
PloS one, 8(12), e84201-e84201 (2014-01-01)
The human dental follicle partially differentiates into the periodontal ligament (PDL), but their biological functions are different. The gene-expression profiles of the dental follicle and PDL were compared using the cDNA microarray technique. Microarray analysis identified 490 genes with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service