Skip to Content
Merck
All Photos(3)

Key Documents

HPA009654

Sigma-Aldrich

Anti-THRA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-erbA-alpha, Anti-EAR-7, Anti-EAR7, Anti-Nuclear receptor subfamily 1 group A member 1, Anti-Thyroid hormone receptor alpha, Anti-c-erbA-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AFEHYVNHRKHNIPHFWPKLLMKEREVQSSILYKGAAAEGRPGGSLGVHPEGQQLLGMHVVQGPQVRQLEQQLGEAGSLQGPVLQHQSPKSPQQRLLELLHRSGILHARAVCGEDDSSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THRA(7067)

General description

THRA (thyroid hormone receptor α) belongs to a family of nuclear receptors, which function as ligand-regulated transcription factors and are called thyroid hormone receptors (TRs). THRA is a predominant mammalian TR isoform. This receptor is highly expressed in central nervous system, myocardium, gastrointestinal tract and skeletal muscle.

Immunogen

Thyroid hormone receptor alpha recombinant protein epitope signature tag (PrEST)

Application

Anti-THRA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Conserved lysine in the DNA-binding domain of this receptor functions as a sensor of transcriptional regulation. This lysine is mutated in hepatocellular carcinoma (HCC). Mutations in this protein result in resistance to thyroid hormone, which is alleviated by thyroxine treatment. Population with rs939348 TT polymorphism in this gene show weak relation to increased risk to Alzheimer′s disease. This receptor is also up-regulated in patients with breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86688

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla Moran et al.
The lancet. Diabetes & endocrinology, 2(8), 619-626 (2014-06-28)
The thyroid hormone receptor α gene (THRA) transcript is alternatively spliced to generate either thyroid hormone receptor (TR)α1 or a non-hormone-binding variant protein, TRα2, the function of which is unknown. Here, we describe the first patients identified with a mutation
Katarzyna J Jerzak et al.
Breast cancer research and treatment, 149(1), 293-301 (2014-12-30)
We determined the expression of two transcriptional variants of thyroid hormone receptor alpha (THRα1 and THRα2) in samples from a cohort of breast cancer patients and correlated expression levels with survival. 130 women who were diagnosed with invasive breast carcinoma
Ivan H Chan et al.
Molecular cancer research : MCR, 8(1), 15-23 (2010-01-08)
Nuclear receptors are hormone-regulated transcription factors that play key roles in normal physiology and development; conversely, mutant nuclear receptors are associated with a wide variety of neoplastic and endocrine disorders. Typically, these receptor mutants function as dominant negatives and can
Louisa Goumidi et al.
Neurobiology of aging, 32(4), 624-630 (2009-05-12)
Because the action of thyroid hormone (T3) is involved in adult cognitive functions, we wanted to assess the association between THRA gene polymorphisms, which encodes the T3 nuclear receptor TRα1, and Alzheimer's disease (AD) risk. We analysed 5 single nucleotide
Erik Björling et al.
Molecular & cellular proteomics : MCP, 7(10), 2028-2037 (2008-08-01)
Antibodies are useful tools to characterize the components of the human proteome and to validate potential protein biomarkers discovered through various clinical proteomics efforts. The lack of validation results across various applications for most antibodies often makes it necessary to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service