Skip to Content
Merck
All Photos(6)

Key Documents

HPA008928

Sigma-Aldrich

Anti-FCER2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BLAST-2, Anti-CD23 antigen, Anti-Fc-epsilon-RII, Anti-Immunoglobulin E-binding factor, Anti-Low affinity immunoglobulin epsilon Fc receptor, Anti-Lymphocyte IgE receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VHARYACDDMEGQLVSIHSPEEQDFLTKHASHTGSWIGLRNLDLKGEFIWVDGSHVDYSNWAPGEPTSRSQGEDCVMMRGSGRWNDAFCDRKLGAWVCDRLATCTPPASEGSAESMGPDSRPDP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCER2(2208)

General description

FCER2 (Fc fragment of IgE receptor II) gene is localized to 19p13.3, spans ~13kb, and is composed of 11 exons. The encoded protein is a low-affinity glycoprotein receptor for immunoglobulin E (IgE). This protein is composed of a single chain and has a molecular weight of 36kDa. It shares high homology with multiple lectins. It exists as two isoforms due to alternative splicing, which differ in their cytoplasmic N-terminals. It is also commonly known as CD23 (cluster of differentiation 23).

Immunogen

Low affinity immunoglobulin epsilon Fc receptor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FCER2 (Fc fragment of IgE receptor II) gene is thought to have regulatory functions in atopic disorders based on its chromosomal location. This receptor controls the production of IgE on B-cells. It is dependent on Ca2+, which causes structural changes in FCER2, which leads to an increased affinity of FCER2 to immunoglobulin (Ig)E. Heightened levels of serum CD23 is linked with increased risk of non-Hodgkin lymphoma. Studies in Xakriabá indigenous community from Brazil shows that the immune-dependent release of nitric oxide (NO) in a CD23-IgE-mediated manner is a hallmark of patients with localized cutaneous leishmaniasis (LCL).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71729

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daopeng Yuan et al.
The Journal of biological chemistry, 288(30), 21667-21677 (2013-06-19)
Immunoglobulin E (IgE) antibodies play a fundamental role in allergic disease and are a target for therapeutic intervention. IgE functions principally through two receptors, FcεRI and CD23 (FcεRII). Minute amounts of allergen trigger mast cell or basophil degranulation by cross-linking
R Carvalho-Gontijo et al.
Scandinavian journal of immunology, 81(6), 515-524 (2015-03-25)
In this study, we described, for the first time, specific aspects of an anti-Leishmania immune response in a Brazilian Xakriabá indigenous community. Induction of an intracellular NO pathway, triggered by the binding of IgE to CD23 receptor in IFN-γ/IL-4 cytokines
Elevated serum sCD23 and sCD30 up to two decades prior to diagnosis associated with increased risk of non-Hodgkin lymphoma.
M P Purdue et al.
Leukemia, 29(6), 1429-1431 (2015-01-09)
T Laitinen et al.
American journal of respiratory and critical care medicine, 161(3 Pt 1), 700-706 (2000-03-11)
On the basis of studies with animal models, the gene for the low-affinity receptor for immunoglobulin E (IgE) (FCER2, CD23) has been implicated as a candidate for IgE-mediated allergic diseases and bronchial hyperreactivity, or related traits. Given evidence for genetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service