Skip to Content
Merck
All Photos(7)

Key Documents

HPA008750

Sigma-Aldrich

Anti-CD247 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD247 antigen, Anti-T-cell receptor T3 zeta chain, Anti-T-cell surface glycoprotein CD3 zeta chain precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KFSRSADAPAYQQGQNQLYNELNLGRREEYDVLDKRRGRDPEMGGKPRRKNPQEGLYNELQKDKMAEAYSEIGMKGERRRGKGHDGLYQGLSTATKDTYDALHMQALP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD247(919)

General description

CD247 (cluster of differentiation 247), also known as T-cell receptor T3 zeta chain (CD3ξ), forms a part of the TCR (T-cell receptor)-CD3 complex. This gene is localized to human chromosome 1q22-q23 and is composed of eight exons. It is one of the four invariant subunits of TCR-CD3 complex.

Immunogen

T-cell surface glycoprotein CD3 zeta chain precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CD247 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

CD247 (cluster of differentiation 247), also known as T-cell receptor T3 zeta chain (CDζ), plays an essential role in the organization, surface expression and signaling pathway of T-cell receptor (TCR)-CD3 complex. Any aberrations in these processes result in T-cell dysfunction and autoimmune disorder. Polymorphisms in this gene are linked to systemic lupus erythematosus. Variants in this gene are also linked to susceptibility to rheumatoid arthritis. In chronic hepatitis C patients, myeloid-derived suppressor cells (MDSCs) lead to persistence of viral infection and suppression of CD247 expression on CD8+ T-cells. Thus, MDSCs lead to dysfunction of T-cells mediated through CD247. Polymorphisms of this gene influence the response to hepatitis B vaccination, which in turn is determined by the BMI (body mass index).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70699

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsutomu Takeuchi et al.
Rheumatology (Oxford, England), 52(9), 1551-1555 (2013-03-26)
SLE is associated with a deficiency in cluster of differentiation 247 (CD247, also known as CD3 zeta chain), a component of the T-cell receptor (TCR)-CD3 complex. A comprehensive analysis showed that in more than half of SLE patients tested CD247
Seamus Grundy et al.
PloS one, 8(8), e71629-e71629 (2013-08-27)
CD8 cells may contribute towards an autoimmune process in COPD. Down regulation of T cell receptor (TCR) signalling molecules occurs in autoimmune diseases with consequent T cell dysfunction. We hypothesise that TCR signalling is abnormal in COPD pulmonary CD8 cells.
Wei Xia Ang et al.
Oncotarget, 8(8), 13545-13559 (2017-01-16)
The epithelial cell adhesion molecule (EpCAM) is overexpressed in a wide variety of tumor types, including peritoneal carcinomatosis (PC) from gastrointestinal and gynecological malignancies. To develop a chimeric antigen receptor T (CART) cell therapy approach to treat patients with end-stage
Qing-Lei Zeng et al.
Molecules and cells, 37(1), 66-73 (2014-02-21)
Myeloid-derived suppressor cells (MDSCs) play an important role in impairing the function of T cells. We characterized MDSCs in two chronic hepatitis C (CHC) cohorts: a cross-sectional group that included 61 treatment-naive patients with CHC, 14 rapid virologic response (RVR)
María Teruel et al.
PloS one, 8(7), e68295-e68295 (2013-07-19)
Given the role of CD247 in the response of the T cells, its entailment in autoimmune diseases and in order to better clarify the role of this gene in RA susceptibility, we aimed to analyze CD247 gene variants previously associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service