Skip to Content
Merck
All Photos(7)

Key Documents

HPA008246

Sigma-Aldrich

Anti-MUC5B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cervical mucin, Anti-High molecular weight salivary mucin MG1, Anti-MUC-5B, Anti-Mucin-5 subtype B, tracheobronchial, Anti-Mucin-5B precursor, Anti-Sublingual gland mucin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

CTCTYEDRTYSYQDVIYNTTDGLGACLIAICGSNGTIIRKAVACPGTPATTPFTFTTAWVPHSTTSPALPVSTVCVREVCRWSSWYNGHRPEPGLGGGDFETFENLRQRGYQVCPVLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC5B(727897)

General description

Mucin 5B (MUC5B) gene is mapped to human chromosome 11p15.5. It codes for a 3570 amino acid peptide characterized with a Cys-subdomains of 108 amino acids.

Immunogen

Mucin-5B precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-MUC5B antibody produced in rabbit has been used in immunofluorescence

Biochem/physiol Actions

Mucin 5B (MUC5B) plays a vital role in maintaining immune homeostasis in the lungs by facilitating mucociliary clearance (MCC). Mucins are the key components that imparts gel-like properties to the mucus.(7) Mutation in the gene leads to the development of idiopathic pulmonary fibrosis. Upregulated expression of MUC5B in the ear causes middle ear effusions.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70004

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muc5b Is Required for Airway Defense
Roy MG, et al.
Nature, 505, 412-412 (2014)
Characterization of an immortalized human small airway basal stem/progenitor cell line with airway region-specific differentiation capacity
Wang G, et al.
Respiratory Research, 20, 196-196 (2019)
Katharina Ernst et al.
Scientific reports, 11(1), 5429-5429 (2021-03-10)
Whooping cough is caused by Bordetella pertussis that releases pertussis toxin (PT) which comprises enzyme A-subunit PTS1 and binding/transport B-subunit. After receptor-mediated endocytosis, PT reaches the endoplasmic reticulum from where unfolded PTS1 is transported to the cytosol. PTS1 ADP-ribosylates G-protein
Nina Lodes et al.
Tissue engineering. Part A, 26(7-8), 432-440 (2019-11-08)
Three-dimensional respiratory tissue models have been generated using, for example, human primary airway epithelial cells (hAEC) or respective cell lines. To investigate ciliopathies, such as primary ciliary dyskinesia, the presence of functional kinocilia in vitro is an essential prerequisite. Since
A Common MUC5B Promoter Polymorphism and Pulmonary Fibrosis
Seibold MA, et al.
The New England Journal of Medicine, 364, 1503-1512 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service