Skip to Content
Merck
All Photos(8)

Key Documents

HPA007308

Sigma-Aldrich

Anti-NQO1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Azoreductase antibody produced in rabbit, Anti-DT-diaphorase antibody produced in rabbit, Anti-DTD antibody produced in rabbit, Anti-Menadione reductase antibody produced in rabbit, Anti-NAD(P)H dehydrogenase [quinone] 1 antibody produced in rabbit, Anti-NAD(P)H:quinone oxidoreductase 1 antibody produced in rabbit, Anti-Phylloquinone reductase antibody produced in rabbit, Anti-QR1 antibody produced in rabbit, Anti-Quinone reductase 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FRSKKAVLSITTGGSGSMYSLQGIHGDMNVILWPIQSGILHFCGFQVLEPQLTYSIGHTPADARIQILEGWKKRLENIWDETPLYFAPSSLFDLNFQAGFLMKKEVQDEEKNKKFGLSVGHHLGKSIPTDNQIKAR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NQO1(1728)

General description

NAD(P)H:quinone oxidoreductase 1 is a homodimer that is predominantly cytosolic. Each monomer contains one molecule of FAD. The gene is localized to human chromosome 16q22.1 and is a single copy gene. It consists of six exons interspaced by five introns and spans a length of 20kb. The 5′UT, first two amino acids, and the first nucleotide of the third amino acid are encoded by exon 1. The rest of the exons encode the remaining 272 amino acids and the 3′UT.

Immunogen

NAD(P)H dehydrogenase [quinone] 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-NQO1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

NQO1 (NAD(P)H:quinone oxidoreductase 1) gene encodes an obligate two-electron reductase that utilizes either NADH or NADPH as a reducing cofactor. It catalyzes reduction of substrates such as quinones, quinone-imines, glutathionyl-substituted naphthoquinones, dichlorophenolindolphenol, methylene blue, azo and nitro compounds, dinitropyrenes, nitrophenylaziridines and nitrobenzamides. It also catalyzes four-electron reduction of azo dyes and nitro compounds. It functions in the detoxification of redox-cycling quinones such as menadione. It also plays the role of an antioxidant by regenerating antioxidant forms of ubiquinone and Vitamin E. By catalyzing these reactions, it protects cells from oxidant stress and electrophilic attack. Defects in this gene have been associated with tardive dyskinesia (TD) and an increased risk of hematotological malignancy after exposure to benzene. Polymorphism in this gene also increases the susceptibility to various cancers

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70184

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Yang et al.
Cancer science, 112(2), 641-654 (2020-11-23)
Patients with hepatocellular carcinoma (HCC) are usually diagnosed at the later stages and have poor survival outcomes. New molecules are urgently needed for the prognostic predication and individual treatment. Our study showed that high levels of NQO1 expression frequently exist
Luke M Shelton et al.
Kidney international, 88(6), 1261-1273 (2015-10-01)
The transcription factor Nrf2 exerts protective effects in numerous experimental models of acute kidney injury, and is a promising therapeutic target in chronic kidney disease. To provide a detailed insight into the regulatory roles of Nrf2 in the kidney, we
Chang Jiang et al.
Redox biology, 54, 102358-102358 (2022-06-07)
The redox regulator NRF2 is hyperactivated in a large percentage of non-small cell lung cancer (NSCLC) cases, which is associated with chemotherapy and radiation resistance. To identify redox vulnerabilities for KEAP1/NRF2 mutant NSCLC, we conducted a CRISPR-Cas9-based negative selection screen
Yan Zhang et al.
Cancer investigation, 33(2), 39-40 (2015-01-23)
Numerous studies have investigated the association between NQO1 Pro187Ser polymorphism and urinary system cancer risk, but the findings are inconsistent. To derive a more precise estimation of such association, we performed a meta-analysis based on 22 publications encompassing 5,274 cases
Guillaume Beinse et al.
PloS one, 14(3), e0214416-e0214416 (2019-03-26)
NRF2 is a major transcription factor regulating the expression of antioxidative/detoxifying enzymes, involved in oncogenic processes and drug resistance. We aimed to identify molecular alterations associated with NRF2 activation in endometrial carcinoma (EC). Ninety patients treated (2012-2017) for localized/locally advanced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service