Skip to Content
Merck
All Photos(3)

Key Documents

HPA006692

Sigma-Aldrich

Anti-RAD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HRAD1, Anti-RAD1 homolog (S. pombe), Anti-REC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GLREAFSELDMTSEVLQITMSPDKPYFRLSTFGNAGSSHLDYPKDSDLMEAFHCNQTQVNRYKISLLKPSTKALVLSCKVSIRTDNRGFLSLQYMIRNEDGQICFVEYYCCPDEEVPESES

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD1(5810)

Immunogen

RAD1 checkpoint DNA exonuclease

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAD1 (RAD1 homolog, S. pombe) is a component of Rad9-Rad1-Hus1 complex (9-1-1 complex) highly expressed in the testis. Each protein of 9-1-1 complex has a binding site for holding the other two partners. It plays a key role in cell cycle checkpoint signaling pathways. It is activated by any DNA stress such as DNA damage and incomplete DNA replication. It reflects response at the S-phase and G2/M arrest stage to identify DNA damage. Altered expression of RAD1 have been reported in non-small-cell lung cancer, squamous cell carcinomas, adenocarcinomas, and bladder cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71060

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Burtelow et al.
The Journal of biological chemistry, 276(28), 25903-25909 (2001-05-08)
DNA damage activates cell cycle checkpoint signaling pathways that coordinate cell cycle arrest and DNA repair. Three of the proteins involved in checkpoint signaling, Rad1, Hus1, and Rad9, have been shown to interact by immunoprecipitation and yeast two-hybrid studies. However
H A Bluyssen et al.
Genomics, 54(2), 331-337 (1998-11-26)
The Schizosaccharomyces pombe rad1+ cell cycle checkpoint control gene is required for S-phase and G2/M arrest in response to both DNA damage and incomplete DNA replication. We isolated and characterized the putative human RAD1 (hRAD1) and mouse RAD1 (mRAD1) homologs
U K Marathi et al.
Genomics, 54(2), 344-347 (1998-11-26)
Cell cycle checkpoints are gating mechanisms that govern cell cycle progression in the presence of DNA damage and incomplete DNA replication. The Schizosaccharomyces pombe Rad1 protein is an essential component of cell cycle checkpoints activated by both types of genomic
Cecilia Cotta-Ramusino et al.
Science (New York, N.Y.), 332(6035), 1313-1317 (2011-06-11)
The DNA damage response (DDR) is brought about by a protein kinase cascade that orchestrates DNA repair through transcriptional and posttranslational mechanisms. Cell cycle arrest is a hallmark of the DDR. We screened for cells that lacked damage-induced cell cycle
Åsa Ehlén et al.
Translational oncology, 4(4), 212-221 (2011-08-02)
The RNA-binding motif protein 3 (RBM3) was initially discovered as a putative cancer biomarker based on its differential expression in various cancer forms in the Human Protein Atlas (HPA). We previously reported an association between high expression of RBM3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service