Skip to Content
Merck
All Photos(6)

Key Documents

HPA006660

Sigma-Aldrich

Anti-LMNA antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-70 kDa lamin, Anti-Lamin-A/C, Anti-Renal carcinoma antigen NY-REN-32

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EVVSREVSGIKAAYEAELGDARKTLDSVAKERARLQLELSKVREEFKELKARNTKKEGDLIAAQARLKDLEALLNSKEAALSTALSEKRTLEGELHDLRGQVAKLEAALGEAKKQLQDEMLRRVDAENRLQTMKEELDFQKNIYSEELRE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMNA(4000)

General description

Lamin A/C (LMNA) gene codes for lamin A and lamin C protein. Lamin A is a nuclear intermediate filament protein. The LMNA gene is mapped to human chromosome 1q22.

Immunogen

Lamin-A/C recombinant protein epitope signature tag (PrEST)

Application

Anti-LMNA antibody produced in rabbit has been used in:
  • western blotting (1:100)
  • immunostaining (1:10)
  • indirect immunofluorescence
  • confocal microscopy (1:100)

Biochem/physiol Actions

Lamins play a key role in nuclear stability, chromatin organization, and gene expression. Lamin A (LMNA) participates in the structural organization of the nucleus. It modulates gene expression via interaction with signaling molecules and transcription factors. Different mutations in the LMNA gene results in several genetic disorders called laminopathies. Mutations in the LMNA gene are also associated with muscular dystrophies, neuropathies, dilated cardiomyopathy with conduction disease, lipodystrophies, and premature aging diseases.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70720

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rasha A Al-Saaidi et al.
European journal of heart failure, 20(10), 1404-1412 (2018-06-27)
Lamin A/C mutations are generally believed to be associated with a severe prognosis. The aim of this study was to investigate disease expression in three affected families carrying different LMNA missense mutations. Furthermore, the potential molecular disease mechanisms of the
Karolin Klement et al.
Mechanisms of ageing and development, 133(7), 508-522 (2012-06-26)
Cellular senescence is a permanent cell cycle arrest induced by short telomeres or oncogenic stress in vitro and in vivo. Because no single of the established biomarkers can reliably identify senescent cells, the application of new ones may aid the
Rasha Al-Saaidi et al.
Experimental cell research, 319(19), 3010-3019 (2013-09-05)
Dilated cardiomyopathy (DCM) is a disease of the heart muscle characterized by cardiac chamber enlargement and reduced systolic function of the left ventricle. Mutations in the LMNA gene represent the most frequent known genetic cause of DCM associated with disease
Zhangrong Jia et al.
Medicine, 100(19), e25910-e25910 (2021-06-10)
It is well known that many genetic factors are involved in the occurrence and progression of atrioventricular block (AV block) and atrial fibrillation (AF). However, the genetic variants discovered so far have only explained parts of these processes. More genes
Lu Kong et al.
Carcinogenesis, 33(4), 751-759 (2012-02-04)
Prostate cancer (PC) remains the second most common cause of cancer-related death in Western countries. A previous proteomics study suggested that the nuclear membrane protein lamin A/C to be a maker to discriminate low- and high-Gleason score tumors and to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service